Skip to Content
Merck
All Photos(1)

Key Documents

C0869

Sigma-Aldrich

Celastrol

≥98% (HPLC), solid, antioxidant

Synonym(s):

10-Hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydro-picene-2-carboxylic acid, Tripterin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H38O4
CAS Number:
Molecular Weight:
450.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Celastrol, ≥98% (HPLC), solid

Quality Level

Assay

≥98% (HPLC)

form

solid

color

red

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

CC1=C(O)C(=O)C=C2C1=CC=C3[C@@]2(C)CC[C@@]4(C)[C@@H]5C[C@@](C)(CC[C@]5(C)CC[C@]34C)C(O)=O

InChI

1S/C29H38O4/c1-17-18-7-8-21-27(4,19(18)15-20(30)23(17)31)12-14-29(6)22-16-26(3,24(32)33)10-9-25(22,2)11-13-28(21,29)5/h7-8,15,22,31H,9-14,16H2,1-6H3,(H,32,33)/t22-,25-,26-,27+,28-,29+/m1/s1

InChI key

KQJSQWZMSAGSHN-JJWQIEBTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Celastrol has been:
  • used intraperitoneally to inject into experimental mice to study its role in nonalcoholic fatty liver disease (NAFLD)
  • used in culture medium to study its effect on lipopolysaccharide-induced inflammatory injury in human keratinocyte cell culture
  • used to study its effect in vitro and in vivo in VCaP cells

Biochem/physiol Actions

Celastrol is a potent antioxidant, and anti-inflammatory agent.
Celastrol is a potent antioxidant, and anti-inflammatory agent. It is a novel HSP90 inhibitor (disrupts Hsp90/Cdc37 complex), that exhibits anticancer (anti-angiogenic - suppresses VEGFR expression); antioxidant (inhibits lipid peroxidation) and anti-inflammatory activity (suppresses iNOS and inflammatory cytokine production).
Celastrol is a quinone methide triterpene naturally present in Tripterygium wilfordii. It is useful in treating rheumatoid arthritis. Celastrol prevents the action of proteasome and nuclear factor Κb.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Celastrol ameliorates liver metabolic damage caused by a high-fat diet through Sirt1
Zhang Y, et al.
Molecular Metabolism, 6(1), 138-147 (2017)
A Mariño-Enríquez et al.
Oncogene, 33(14), 1872-1876 (2013-04-16)
Most gastrointestinal stromal tumors (GISTs) contain KIT or PDGFRA kinase gain-of-function mutations, and therefore respond clinically to imatinib and other tyrosine kinase inhibitor (TKI) therapies. However, clinical progression subsequently results from selection of TKI-resistant clones, typically containing secondary mutations in
Radhamani Kannaiyan et al.
Cancer letters, 303(1), 9-20 (2010-12-21)
Identification of active constituents and their molecular targets from traditional medicine is an enormous opportunity for modern pharmacology. Celastrol is one such compound that was originally identified from traditional Chinese medicine (Thunder of God Vine) almost three decades ago and
A C Allison et al.
Progress in neuro-psychopharmacology & biological psychiatry, 25(7), 1341-1357 (2001-08-22)
In the brains of patients with Alzheimer's disease (AD) signs of neuronal degeneration are accompanied by markers of microglial activation, inflammation, and oxidant damage. The presence of nitrotyrosine in the cell bodies of neurons in AD suggests that peroxynitrite contributes
Tripterine alleviates LPS-induced inflammatory injury by up-regulation of miR-146a in HaCaT cells
Xiong Y, et al.
Biomedicine and Pharmacotherapy, 105, 798-804 (2018)

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service