Skip to Content
Merck
All Photos(1)

Key Documents

Y0002055

Fingolimod hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

FTY720, 2-Amino-2-[2-(4-octyl-phenyl)-ethyl]-propane-1,3-diol hydrochloride, Fingolimod hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H33NO2 · HCl
CAS Number:
Molecular Weight:
343.93
UNSPSC Code:
41116107
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

fingolimod

form

powder

packaging

pkg of 110 mg

manufacturer/tradename

EDQM

storage condition

protect from light

solubility

water: 100-1000 g/L

application(s)

pharmaceutical

format

neat

shipped in

ambient

storage temp.

2-8°C

SMILES string

Cl.NC(CO)(CO)CCc1ccc(cc1)CCCCCCCC

InChI

1S/C19H33NO2.ClH/c1-2-3-4-5-6-7-8-17-9-11-18(12-10-17)13-14-19(20,15-21)16-22;/h9-12,21-22H,2-8,13-16,20H2,1H3;1H

InChI key

SWZTYAVBMYWFGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fingolimod Hydrochloride (fingolimod HCl), an S1P-receptor modulator, belongs to the class of synthetic glucocorticoid steroid drugs.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

FTY720 is an immunomodulating drug and sphingosine 1-phosphate (S1P) receptor modulator. Phosphorylation of FTY270 by sphingosine kinase causes S1P1R internalization, which sequesters lymphocytes in lymph nodes, preventing them from taking part in an autoimmune response. Clinically, it has been approved for the treatment of multiple sclerosis (MS). It has also been shown to block and reverse paclitaxel-induced chemotherapy induced peripheral neuropathy (CIPN) through S1PR inhibition as well as inhibit the activity of histone deacetylases in the hippocampus of mouse brains, thereby modulating memory.
FTY720, an immunomodulating drug, acts as a sphingosine 1-phosphate (S1P) receptor modulator.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Target Organs

Immune system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fingolimod Hydrochloride
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 4(1), 4989-4990 (2019)
A randomized, open-label study assessing the bioequivalence of two formulations of Fingolimod 0.5 mg in healthy subjects
Tanguay M, et al.
AAPS Open, 4(1), 3-3 (2018)
Rapid determination of fingolimod hydrochloride-related Substances and degradation products in API and pharmaceutical dosage forms by use of a stability-indicating UPLC method
Rajan N and Basha KA
Chromatographia, 77(21-22), 1545-1552 (2014)
Sphingosine 1-phosphate receptor 1 as a useful target for treatment of multiple sclerosis
Chiba K and Adachi K
Pharmaceuticals (Basel, Switzerland), 5(5), 514-528 (2012)
Fingolimod hydrochloride gel for dermatological applications: optimization of formulation strength and effect of colloidal oatmeal (Aveeno?) as penetration enhancer
Tamakuwala M and Stagni G
Aaps Pharmscitech, 17(4), 907-914 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service