Skip to Content
Merck
All Photos(3)

Key Documents

C0678

Sigma-Aldrich

Monoclonal Anti-HNK-1/N-CAM (CD57) antibody produced in mouse

clone VC1.1, ascites fluid, buffered aqueous solution

Synonym(s):

Monoclonal Anti-HNK-1, Anti-CD57, Anti-N-CAM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

VC1.1, monoclonal

form

buffered aqueous solution

contains

15 mM sodium azide

species reactivity

mouse, rat, feline, human

technique(s)

western blot: 1:4,000 using cerebral cortex extract

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-HNK-1/N-CAM (CD57) (mouse IgM isotype) is derived from the VC1.1 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. Human natural killer cell -1 (HNK-1), also referred as B3GAT1, NK1, CD57, LEU7, NK-1, GLCATP and GLCUATP, is mapped to human chromosome 11q25. The encoded protein belongs to the glucuronyltransferase gene family. Neural cell-adhesion molecule (N-CAM) is expressed on all neuroepithelial cells from the time of neural induction in the early embryo. It is known that the sulfated carbohydrate HNK-1 epitope is carried by several surface molecules including a proportion of N-CAM isoforms.

Specificity

Recognizes the CD57/HNK-1 human myeloid cell associated surface glycoprotein. The epitope recognized is an N-linked carbohydrate which is present in a variety of glycoproteins and in some glycolipids. It is resistant to formalin fixation and paraffin embedding. VC1.1 antibody and the HNK-1 (Leu7) antibody inhibit the binding of each other. The antibody recognizes myelin associated glycoprotein in some species and a high molecular weight chondroitin sulphate proteoglycan.
5th Workshop: code no. NK 67

Immunogen

A homogenate of cat primary visual cortex (area 17)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Monoclonal Anti-HNK-1/N-CAM (CD57) antibody produced in mouse has been used:
  • as primary antibody for western blot analysis
  • immunocytochemistry
  • immunocytochemical
  • immunohistological staining

Biochem/physiol Actions

HNK1 (human natural killer-1) is referred as B3GAT1, NK1, CD57, LEU7, NK-1, GLCATP and GLCUATP. It is a member of the glucuronyltransferase gene family. Human natural killer-1 (HNK1) carbohydrate epitope is expressed on various cell adhesion molecules in the nervous system and plays a role in cell-cell and cell-substrate interactions. HNK-1 epitope stabilizes GluR2 on neuronal surface membranes and regulates cell surface stability of GluR2 by modulating the interaction with N-cadherin. It is also plays a role in cell migration and synaptic plasticity. HNK-1 epitope has been associated with the risk of metastasis. It is observed to be highly expressed in melanoma cell lines.

Target description

CD57/HNK-1 is expressed on a subpopulation of 15-20% of peripheral blood mononuclear cells, about 60% of NK active cells and on a subset of T cells.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald S Goldstein
Methods in molecular biology (Clifton, N.J.), 331, 137-151 (2006-08-03)
The traditional methods of studying the differentiation of human embryonic stem cells (hESCs) are to differentiate them in vitro or in immune-deficient mice as teratomas. The chick embryo is a well-studied and accessible experimental system that has been shown to
Jean-Marie Gasc et al.
Biology open, 4(5), 666-671 (2015-04-04)
The enteric nervous system originates from neural crest cells that migrate in chains as they colonize the embryonic gut, eventually forming the myenteric and submucosal plexus. Failure of the neural crest cells to colonize the gut leads to aganglionosis in
Yi-Sen Cheng et al.
Anatomical record (Hoboken, N.J. : 2007), 298(7), 1271-1281 (2015-03-27)
Sorafenib has been used as an oral anti-cancer drug because of its ability to inhibit tumor growth. However, the pharmacological effect of sorafenib is still the lack of in vivo experimental evidence. Tumor and embryonic cells share some similar features
Neural differentiation of pluripotent mouse embryonal carcinoma cells by retinoic acid: inhibitory effect of serum
Pachernik J, et al.
Physiological Research, 54(1), 115-122 (2005)
Loss of Shp2-mediated mitogen-activated protein kinase signaling in Muller glial cells results in retinal degeneration
Cai Z, et al.
Molecular and Cellular Biology, 31(14), 2973-2983 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service