Skip to Content
Merck
All Photos(2)

Documents

B7151

Sigma-Aldrich

Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:200,000-1:300,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200
western blot: 1:400,000-1:800,000 using total cell extract of HeLa cells

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG (immunoglobulin G) antibody subtype is a predominant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids. IgG is found in four subclasses IgG1, IgG2, IgG3 and IgG4. IgG has a monomeric structure.
The product binds to all mouse Igs

Application

Anti-Mouse IgG (Fab specific)-Biotin antibody produced in goat was used in immunohistochemistry of lateral and medial menisci of rabbits and human adipose-derived stem cells.

Biochem/physiol Actions

IgG (immunoglobulin G) antibody participates in complement fixation and opsonization.
IgG antibody provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of biotin to Anti-Mouse IgG (Fab specific) antibody allows for the binding of various labels such as avidin or streptavidin.

Other Notes

Antibody adsorbed with bovine, equine and human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with bovine, horse or human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph F Poduslo et al.
Journal of neurochemistry, 102(2), 420-433 (2007-06-29)
Targeting therapeutic or diagnostic proteins to the nervous system is limited by the presence of the blood-brain barrier. We report that a F(ab')(2) fragment of a monoclonal antibody against fibrillar human Abeta42 that is polyamine (p)-modified has increased permeability at
Brian O Diekman et al.
Journal of biomedical materials research. Part A, 93(3), 994-1003 (2009-09-02)
Adipose-derived stem cells (ASCs) are multipotent progenitors that can be chondrogenically induced by growth factors such as bone morphogenetic protein 6 (BMP-6). We hypothesized that nonviral transfection of a BMP-6 construct (pcDNA3-BMP6) would induce chondrogenic differentiation of ASCs encapsulated in
G M Cunniffe et al.
European cells & materials, 33, 130-142 (2017-02-16)
Limitations associated with demineralised bone matrix and other grafting materials have motivated the development of alternative strategies to enhance the repair of large bone defects. The growth plate (GP) of developing limbs contain a plethora of growth factors and matrix
Franklin T Moutos et al.
Tissue engineering. Part A, 16(4), 1291-1301 (2009-11-12)
Articular cartilage possesses complex mechanical properties that provide healthy joints the ability to bear repeated loads and maintain smooth articulating surfaces over an entire lifetime. In this study, we utilized a fiber-reinforced composite scaffold designed to mimic the anisotropic, nonlinear
Franklin T Moutos et al.
Macromolecular bioscience, 10(11), 1355-1364 (2010-09-22)
The successful replacement of large-scale cartilage defects or osteoarthritic lesions using tissue-engineering approaches will likely require composite biomaterial scaffolds that have biomimetic mechanical properties and can provide cell-instructive cues to control the growth and differentiation of embedded stem or progenitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service