Skip to Content
Merck
All Photos(1)

Key Documents

B9806

Sigma-Aldrich

Heparin−biotin sodium salt

≥97%

Synonym(s):

Heparin-Biotin Conjugate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
eCl@ss:
34058011
NACRES:
NA.25

conjugate

biotin conjugate

Quality Level

Assay

≥97%

form

solid

mol wt

~15000 Da

solubility

H2O: soluble 1 mg/mL, clear, colorless

storage temp.

2-8°C

Application

Heparin-biotin is used transfection research for the development of self-assembled terplexes such as polyamidoamine dendrimer and DNA (PAMAM/DNA) heparin-biotin terplexes for targeted gene delivery with improved transfection. The terplexes may be used in immobilizing proteins such as lactoferrin and lysozyme and nucleic acids.

Other Notes

Biotin conjugated to heparin from porcine intestinal mucosa

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Liu et al.
Die Pharmazie, 67(2), 174-181 (2012-04-20)
The efficiency and safety of gene delivery vectors were important factors for gene therapy. To enhance gene transfection efficiency and to incorporate biocompatible components to the polyamidoamine (PAMAM) dendrimer mediated gene delivery systems, human serum albumin (HSA) was introduced to
S Zou et al.
Comparative biochemistry and physiology. B, Comparative biochemistry, 103(4), 889-895 (1992-12-01)
1. Binding of biotin-heparin to immobilized lactoferrin and lysozyme was optimum at pH 6.0, 100 mM NaCl. Complex interactions between NaCl and CaCl2 concentrations were observed for heparin binding to both proteins. 2. The metal ions Cu2+, Zn2+, Fe2+ and
Xue-Qing Zhang et al.
Bioconjugate chemistry, 18(6), 2068-2076 (2007-09-13)
We report on the preparation and characterization of poly(D, L-lactide-co-glycolide) (PLGA) microparticles with surface-conjugated polyamidoamine (PAMAM) dendrimers of varying generations. The buffering capacity and zeta-potential of the PLGA PAMAM microparticles increased with increasing generation level of the PAMAM dendrimer conjugated.
Dzmitry Shcharbin et al.
Journal of controlled release : official journal of the Controlled Release Society, 141(2), 110-127 (2009-10-10)
This paper reviews different techniques for analyzing the transfection efficiencies and cytotoxicities of dendriplexes-complexes of nucleic acids with dendrimers. Analysis shows that three plasmids are mainly used in transfection experiments: plasmid DNA encoding luciferase from the firefly Photinus pyralis, beta-galactosidase
Qiao Zhang et al.
Bioconjugate chemistry, 21(11), 2086-2092 (2010-10-12)
To improve transfection efficiency and to incorporate target ligands to the gene delivery systems, heparin and heparin-biotin were introduced to complexes of polyamidoamine dendrimer and DNA (PAMAM/DNA) via electrostatic interactions to form self-assembled PAMAM/DNA/heparin and PAMAM/DNA/heparin-biotin terplexes, respectively. The self-assembled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service