Skip to Content
Merck
All Photos(1)

Documents

B9151

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl butyrate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11BrClNO2
CAS Number:
Molecular Weight:
316.58
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Quality Level

Assay

≥99%

form

powder

solubility

acetone: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CCCC(=O)Oc1c[nH]c2ccc(Br)c(Cl)c12

InChI

1S/C12H11BrClNO2/c1-2-3-10(16)17-9-6-15-8-5-4-7(13)12(14)11(8)9/h4-6,15H,2-3H2,1H3

InChI key

UKTKOBRRRRODGL-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service