Skip to Content
Merck
All Photos(3)

Key Documents

B8684

Sigma-Aldrich

Bambuterol hydrochloride

>98% (HPLC), powder

Synonym(s):

Dimethylcarbmic acid 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-1,3-phenylene ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H29N3O5 · HCl
CAS Number:
Molecular Weight:
403.90
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>98% (HPLC)

form

powder

storage condition

desiccated

color

off-white

solubility

H2O: soluble ~33 mg/mL

originator

AstraZeneca

SMILES string

Cl.CN(C)C(=O)Oc1cc(OC(=O)N(C)C)cc(c1)C(O)CNC(C)(C)C

InChI

1S/C18H29N3O5.ClH/c1-18(2,3)19-11-15(22)12-8-13(25-16(23)20(4)5)10-14(9-12)26-17(24)21(6)7;/h8-10,15,19,22H,11H2,1-7H3;1H

InChI key

LBARATORRVNNQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bambuterol is present in various tissues, like , intestine, lungs, liver and heart. It is also found in serum.

Application

Bambuterol hydrochloride has been used as a β2-adrenergic receptor (β2-AR) agonist to study its effects on graft versus host disease (GVHD) while preserving the graft-versus-tumor (GvT) effect. It has also been used in the preparation of bambuterol-poly vinyl chloride (Bb-PVC) membrane sensors for the determination of bambuterol in the pharmaceutical dosage form.

Biochem/physiol Actions

Bambuterol hydrochloride is used for prophylaxis and also to treat chronic bronchitis and chronic asthma in pediatrics.
β-adrenoceptor agonist; bronchodilator; anti-asthmatic; terbutaline prodrug

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PVC membrane sensors for potentiometric determination of bambuterol in pharmaceutical formulation
Ezzeldin E, et al.
International Journal of Electrochemical Science, 7(11), 10570-10581 (2012)
Yejin Jang et al.
Viruses, 12(5) (2020-05-30)
Influenza A virus, one of the major human respiratory pathogens, is responsible for annual seasonal endemics and unpredictable periodic pandemics. Despite the clinical availability of vaccines and antivirals, the antigenic diversity and drug resistance of this virus makes it a
Hemn Mohammadpour et al.
JCI insight, 5(12) (2020-05-22)
Acute graft versus host disease (aGvHD) remains a major impediment to successful allogeneic hematopoietic cell transplantation (allo-HCT). To solve this problem, a greater knowledge of factors that regulate the differentiation of donor T cells toward cytotoxic cells or Tregs is
beta2-adrenergic receptor activation on donor cells ameliorates acute GvHD
Mohammadpour H, et al.
JCI insight (2020)
J Rosenborg et al.
British journal of clinical pharmacology, 49(3), 199-206 (2000-03-16)
The pharmacokinetics of orally administered bambuterol were investigated in healthy adult subjects, with particular regard to time to steady state, pharmacokinetic linearity, intraindividual variability for the parent drug and its active beta2-adrenergic metabolite terbutaline and bioequivalence between tablet and solution.

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service