Skip to Content
Merck
All Photos(10)

Key Documents

AMAB91130

Sigma-Aldrich

Anti-Choline Acetyltransferase Antibody

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, mouse monoclonal, CL3173

Synonym(s):

Anti-CHOACTASE, Anti-CMS1A, Anti-CMS1A2, Anti-CMS6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

Product Name

Monoclonal Anti-CHAT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL3173, purified immunoglobulin, buffered aqueous glycerol solution

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CL3173, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 2-10 μg/mg (Fixation/Permeabilization: PFA/Triton X-100)
immunohistochemistry: 1:1000- 1:2500

isotype

IgG2b

immunogen sequence

GLFSSYRLPGHTQDTLVAQNSSIMPEPEHVIVACCNQFFVLDVVINFRRLSEGDLFTQLRKIVKMASNEDERLPPIGLLTSDGRSEWAEARTVLVKDSTN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHAT(1103)

General description

Choline O-acetyltransferase (CHAT) gene is mapped to human chromosome 10q11.23. It has a choline binding domain pocket and a catalytic domain.

Immunogen

choline O-acetyltransferase

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Choline O-acetyltransferase (CHAT) enzyme mediates the synthesis of neurotransmitter, acetylcholine from choline and acetyl-CoA in cholinergic neurons. Phosphorylation at serine and threonine residues is critical for the function of CHAT. Mutations in the active site residues, results in loss of enzyme activity. Deletion in CHAT gene locus impacts neuromuscular signal transmission contributing to muscle weakness in congenital myasthenic syndromes. A mutation in the CHAT gene leads to choline acetyltransferase deficiency and triggers recurrent breathlessness in infants. Polymorphisms in CHAT is implicated in Alzheimer′s disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74314

Physical form

40% glycerol and PBS (pH 7.2). 0.02% sodium azide is added as preservative.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of Choline Acetyltransferase Gene Polymorphisms (SNPs rs868750G/A, rs1880676G/A, rs2177369G/A and rs3810950G/A) with Alzheimer?s Disease Risk: A Meta-Analysis
Yuan H, et al.
PLoS ONE, 11(7), e0159022-e0159022 (2016)
Congenital myasthenic syndrome associated with episodic apnea and sudden infant death
Byring RF, et al.
Neuromuscular Disorders, 12(6), 548-553 (2002)
Functional consequences and structural interpretation of mutations of human choline acetyltransferase
Shen Xm, et al.
Human Mutation, 32(11), 1259-1267 (2011)
Linyan Ma et al.
Investigative ophthalmology & visual science, 65(4), 30-30 (2024-04-18)
This study aims to elucidate the calcitonin gene-related peptide (CGRP) mediation and primary mechanism of corneal sensory nerves on tear production of the lacrimal gland. Mouse corneal denervation models were constructed through surgical axotomy, pharmacologic treatment with capsaicin or resiniferatoxin
Congenital myasthenic syndromes: pathogenesis, diagnosis, and treatment
Engel AG, et al.
Lancet Neurology, 14(4), 420-434 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service