Skip to Content
Merck
All Photos(1)

Key Documents

AB0300

Sigma-Aldrich

Alkaline Phosphatase Blue Membrane Substrate Solution

alkaline phosphatase substrate, solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
42010105
NACRES:
NA.83

Product Name

Alkaline Phosphatase Blue Membrane Substrate Solution, sufficient for 400 mL working substrate

form

solution

Quality Level

usage

sufficient for 400 mL working substrate

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Alkaline phosphatase, a metalloenzyme is a member of ectonucleotidases family. It is present in bile ducts, bone, intestine, placenta and tumors.

Application

Alkaline Phosphatase Blue Membrane Substrate Solution has been used:
  • to analyse direct alkaline phosphatase (AP) activity
  • to perform alkaline phosphatase assays with colorimetric system
  • to analyse direct alkaline phosphatase (AP) activity of the induced pluripotent stem cells (iPSC) line LND554SV.3

Prior to reaction with alkaline phosphatase, the BCIP® reagent is colorless to faint blue and the Nitro BT reagent is yellow. The two-component mixture develops a bluish-purple product with alkaline phosphatase in membrane-type assays. This substrate is not recommended for ELISA assays.

Physical form

A 2 component kit sufficient for preparing 400 mL of working substrate.
Supplied as a two-component buffered alkaline phosphatase substrate containing bromochloroindolyl phosphate (BCIP®) and Nitro Blue Tetrazolium (NBT/Thiazolyl Blue/Nitro BT)

Legal Information

BCIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of alkaline phosphatase: an emerging new drug target
Al-Rashida M and Iqbal J
Mini-Reviews in Medicinal Chemistry, 15(1), 41-51 (2015)
The Short Isoform of NF-YA Belongs to the Embryonic Stem Cell Transcription Factor Circuitry
Dolfini D, et al.
Stem Cells, 30(11), 2450-2459 (2012)
Generation of a human iPSC line from a patient with Leigh syndrome
Galera T, et al.
Stem Cell Research, 16(1), 63-66 (2016)
Generation of pig iPS cells: a model for cell therapy
Montserrat N, et al.
Journal of Cardiovascular Translational Research, 4(2), 121-130 (2011)
Jaundice
Nelson Pediatric Symptom-Based Diagnosis, 255-274 (2018)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service