Skip to Content
Merck
All Photos(1)

Documents

A9294

Sigma-Aldrich

Anti-Goat IgG (whole molecule)−Agarose antibody produced in rabbit

IgG fraction of antiserum, PBS suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

agarose conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

PBS suspension

species reactivity

goat

extent of labeling

~2.4 mg Ab per mL

technique(s)

immunoelectrophoresis: suitable

matrix

Cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix spacer

1 atom

capacity

1 mg/mL, resin binding capacity (goat IgG)

storage temp.

2-8°C

target post-translational modification

unmodified

General description

An immunoglobulin has two heavy chains and two light chains connected by disulfide bond. It mainly helps in immune defense. . IgG, a glycoprotein, is the major class of immunoglobulin. Goat IgG has two subclasses- IgG1 and IgG2.
IgGs are glycoprotein antibodies that modulate several immune responses. Goat IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-goat IgGs are useful tools for the analysis of target proteins.

Specificity

Rabbit Anti-Goat IgG (whole molecule)-Agarose antibody binds to goat IgG.

Immunogen

pooled normal goat serum

Application

Anti-Goat IgG (whole molecule) Agarose antibody produced in rabbit has been used in protein purification, immunoprecipitation/chromatin immunoprecipitation assay.
c-RET was immunoprecipitated from overexpressing MDCK cells using agarose-linked rabbit anti-goat IgG. Homogenates were incubated with conjugated beads for 2 hours on ice.

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III.

Physical form

Suspension in phosphate buffered saline containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of a translationally fused TAP-tagged plasma membrane proton pump in Arabidopsis thaliana
Rodrigues RB, et al.
Biochemistry, 53(3) (2014)
An essential role for Orc6 in DNA replication through maintenance of pre-replicative complexes
Semple JW, et al.
The Embo Journal, 25(21), 5150-5158 (2006)
Specific IgG for cat allergens in patients with allergic conjunctivitis
Miyama A, et al.
International Ophthalmology, 575-586 (2015)
Mark W Barnett et al.
Journal of cell science, 115(Pt 23), 4495-4503 (2002-11-05)
Glial cell line-derived neurotrophic factor, GDNF, is vital to the development and maintenance of neural tissues; it promotes survival of sympathetic, parasympathetic and spinal motor neurons during development, protects midbrain dopaminergic neurons from apoptosis well enough to be a promising
Janina Borgonovo et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 30(4), 333-341 (2012-02-07)
It is well known that clathrin-mediated endocytosis is crucial for the normal functioning and integrity of neurons in the central nervous system. In this study we attempted to correlate the expression of coat proteins with development in different areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service