A6582
Antifoam A concentrate
active silicone polymer 100%
Synonym(s):
Antifoam agent, Antifoam solution, Foam control agent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161901
NACRES:
NA.85
Recommended Products
Quality Level
form
emulsion (aqueous)
technique(s)
cell culture | hybridoma: suitable
microbiological culture: suitable
density
0.97 g/mL at 25 °C
application(s)
microbiology
storage temp.
room temp
suitability
(Mammalian (suspension))
bacteria (fermentation)
General description
Antifoam A is a silicone-based product used to prevent or minimize foaming in both microbial fermentation systems and mammalian suspension cultures. Control of foaming minimizes damage to the cells, resulting in increased protein/antibody production. Extremely effective for aqueous and non-aqueous systems. Effective at concentrations in the 1-50ppm range.
Application
Antifoam A concentrate has been used:
- in bioreactor culture for Staphylococcal enterotoxin C (SEC) production.
- to prevent foam formation during the aerobic treatment of sludge.
- to study the impact of surface tension on bioaerosol generation during bubbling.
Features and Benefits
- Molecular biology grade and tested for use in bacterial fermentation.
- Extremely effective foam suppressor for aqueous and non-aqueous systems.
- Made of 100% active silicone polymer, free of emulsifiers.
- Typically, effective at 1-100 ppm.
- Product will be stable in the pH range of 5 to 9.
- Can be directly added to a fermentation medium but it is not recommended that it be pumped to a fermenter on an as-needed basis.
Components
Antifoam A is a 100% active silicon polymer containing no non-ionic emulsifiers .
Preparation Note
Antifoam A can be pre-diluted with 3-10 parts of aqueous propylene glycol or non-aqueous vegetable oil with slow mixing.
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>213.8 °F - closed cup
Flash Point(C)
> 101 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The biodegradability of aquatic worm predated waste activated sludge: A sequential aerobic and anaerobic treatment approach
de Valk S, et al.
bioresource technology reports, 12.0, 100606-100606 (2020)
Bioaerosol Release from Concentrated Microbial Suspensions in Bubbling Processes
Kruglyakova E, et al.
Air quality, atmosphere, & health, 13.0, 2029-2029 (2022)
Experience with a perfo-suction system for the recovery of schistosomes from laboratory rodents
T. F. H. G. Jackson, C. D. Dettman, et al.
Laboratory Animal Science, 16, 65-67 (1982)
Factors affecting staphylococcal enterotoxin C bovine production in milk
Hunt K, et al.
International dairy journal, 39(1), 41-46 (2014)
F P Ernani et al.
Infection and immunity, 61(6), 2596-2601 (1993-06-01)
Substantial evidence indicates that molecules released by infectious organisms affect virulence and influence immunity to infection. The characterization of extracellular molecules and their mechanism of release is therefore critical to understanding host-parasite relationships. The cestode parasite Mesocestoides corti is known
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service