Skip to Content
Merck
All Photos(3)

Key Documents

A5222

Sigma-Aldrich

D-Amino Acid Oxidase from porcine kidney

powder, ≥1.5 units/mg solid

Synonym(s):

D-AAO, D-Amino acid: oxygen oxidoreductase (deaminating)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

Quality Level

specific activity

≥1.5 units/mg solid

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

D-amino acid oxidase (DAO) is a potential target molecule for the treatment of chronic pain and schizophrenia . D-amino acid oxidase is used to oxidize D-amino acids to their corresponding imino acids.

Unit Definition

One unit will oxidatively deaminate 1.0 μmole of D-alanine to pyruvate per minute at pH 8.3 at 25 °C, in the presence of a catalase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Lu Huang et al.
Amino acids, 43(5), 1905-1918 (2012-09-22)
D-Amino acid oxidase (DAAO), a FAD-dependent peroxisomal flavoenzyme that catalyzes oxidation of D-amino acids to hydrogen peroxide, is distributed in the spinal cord almost exclusively expressed within astrocytes. The present study aims to explore potential contributions of spinal DAAO to
Norihiro Shibuya et al.
Nature communications, 4, 1366-1366 (2013-01-24)
In eukaryotes, hydrogen sulphide acts as a signalling molecule and cytoprotectant. Hydrogen sulphide is known to be produced from L-cysteine by cystathionine β-synthase, cystathionine γ-lyase and 3-mercaptopyruvate sulfurtransferase coupled with cysteine aminotransferase. Here we report an additional biosynthetic pathway for
Natsuki Matsumoto et al.
Methods in molecular biology (Clifton, N.J.), 1887, 23-31 (2018-12-07)
Fermented foods produced using edible microorganisms have been consumed worldwide since ancient times. Microorganisms produce some healthy functional substances. Recently, many people have been attracted to the notion that D-body amino acids have healthy functional effects. D-body amino acids convert
R M Abou El-Magd et al.
Journal of psychopharmacology (Oxford, England), 24(7), 1055-1067 (2009-03-31)
D-Amino acid oxidase (DAO) has been established to be involved in the oxidation of D-serine, an allosteric activator of the N-methyl-D-aspartate-type glutamate receptor in the brain, and to be associated with the onset of schizophrenia. The effect of risperidone, a
Cindy J Lee et al.
Metabolites, 12(9) (2022-09-24)
The pancreatic islets of Langerhans are clusters of cells that function as endocrine units synthesizing and releasing insulin and a range of additional peptide hormones. The structural and chemical characteristics of islets change during type 2 diabetes development. Although a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service