Skip to Content
Merck
All Photos(3)

Documents

A2859

Sigma-Aldrich

Monoclonal Anti-ATG5 antibody produced in mouse

clone ATG5-18, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-APG5, Anti-APG5L, Anti-ASP, Anti-ATG5 autophagy related 5 homolog, Anti-hAPG5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ATG5-18, monoclonal

form

buffered aqueous solution

mol wt

antigen ~56 kDa

species reactivity

human, mouse, rat

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: 2.5-5.0 μg/mL using whole extract of human K562 cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATG5(9474)
mouse ... Atg5(11793)
rat ... Atg5(365601)

General description

Monoclonal Anti-ATG5 (mouse IgG2a isotype) is derived from the hybridoma ATG5-18 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with human Atg5, conjugated to keyhole limpet hemocyanin (KLH). The autophagy related 5 (ATG5) gene is mapped to human chromosome 6q21 and encodes a 32 kDa protein.

Specificity

Mouse monoclonal anti-ATG5 antibody reacts with human, rat, and mouse Atg5-Atg12 complex by immunoblotting (∼56kDa) and immunoprecipitation.

Application

Monoclonal Anti-ATG5 antibody produced in mouse has been used in western blotting and immunoprecipitation
Omegasomes were shown to localize to autophagosomes by immunohistochemistry using rabbit anti-ATG5 antibody.

Biochem/physiol Actions

Atg5 (also known as Apg5) is a 32kDa protein that is required for mediating autophagy. Studies have reported that the Atg12-Atg5-Atg16 complex localizes to autophagosome precursors and plays an important role in autophagosome formation. Furthermore, a fragment of Atg5 fragment produced by calpain cleavage is known to have pro-apoptotic properties.
Autophagy related 5 (ATG5), also known as Apg5, is essential for autophagy, a major pathway for bulk degradation of cytoplasmic constituents and organelles. In this process, portions of the cytoplasm are sequestered into double membrane vesicles, the autophagosomes, and subsequently delivered to the lysosome for degradation and recycling. Autophagy is required for normal turnover of cellular components during starvation. It plays an essential role in cellular differentiation, cell death and aging. Defective autophagy may contribute to certain human diseases such as cancer, neurodegenerative diseases, muscular disorders and pathogen infections. Autophagy is an evolutionary conserved pathway seen in all eukaryotic cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allan Sauvat et al.
EBioMedicine, 30, 261-272 (2018-04-03)
Depending on the length of their carbon backbone and their saturation status, natural fatty acids have rather distinct biological effects. Thus, longevity of model organisms is increased by extra supply of the most abundant natural cis-unsaturated fatty acid, oleic acid
Xian Jiang et al.
Molecular cell, 63(2), 229-239 (2016-07-23)
In response to apoptotic stimuli, mitochondria in mammalian cells release cytochrome c and other apoptogenic proteins, leading to the subsequent activation of caspases and apoptotic cell death. This process is promoted by the pro-apoptotic members of the Bcl-2 family of
Mireia Niso-Santano et al.
The EMBO journal, 34(8), 1025-1041 (2015-01-15)
To obtain mechanistic insights into the cross talk between lipolysis and autophagy, two key metabolic responses to starvation, we screened the autophagy-inducing potential of a panel of fatty acids in human cancer cells. Both saturated and unsaturated fatty acids such
Shida Yousefi et al.
Nature cell biology, 8(10), 1124-1132 (2006-09-26)
Autophagy-related gene (Atg) 5 is a gene product required for the formation of autophagosomes. Here, we report that Atg5, in addition to the promotion of autophagy, enhances susceptibility towards apoptotic stimuli. Enforced expression of Atg5-sensitized tumour cells to anticancer drug
Sujan Piya et al.
Blood, 128(9), 1260-1269 (2016-06-09)
Autophagy is a cellular adaptive mechanism to stress, including that induced by chemotherapeutic agents. Reverse phase protein array suggested that high expression of the essential autophagy-related protein, Atg7, was associated with shorter remission in newly diagnosed acute myeloid leukemia (AML)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service