Skip to Content
Merck
All Photos(1)

Key Documents

A2064

Sigma-Aldrich

Anti-Human IgG−Alkaline Phosphatase antibody, Mouse monoclonal

clone GG-5, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Human IgG (γ-chain specific)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

alkaline phosphatase conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

GG-5, monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

direct ELISA: 1:50,000
dot blot: 1:80,000
immunohistochemistry: suitable
western blot (chemiluminescent): 1:80,000

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Binds human IgG; does not bind other human Igs.The antibody is specific for a determinant on the heavy chain of human IgG. This antibody will form antigen-antibody complexes in the liquid phase in the presence of 3% PEG 6000.
Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders.
Monoclonal Anti-Human IgG (Γ-chain specific) (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.

Application

Anti-Human IgG- Alkaline Phosphatase antibody, Mouse monoclonal has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • western blotting
  • dot blot
  • immunohistology

Elisa assays were performed using alkaline phosphatase conjugated monoclonal mouse anti-human IgG to detect antibodies against various S. Aureus antigens in the serum from healthy individuals. The antibody was diluted 1:30000 in PBSt and incubated with samples for 2 hours at 37 degrees. Reactin was developed using p-nitrophenyl phosphate substrate (Sigma).
Liquid precipitate (Lppt).

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 1% bovine serum albumin, 50% glycerol, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George J Broze
Blood cells, molecules & diseases, 52(2-3), 116-120 (2013-10-01)
Acquired factor X (FX) deficiency unrelated to amyloidosis is a rare disorder in which an anti-FX antibody is infrequently detected. A patient with severe bleeding due to a calcium ion-dependent anti-FX IgG antibody is described. The FX affinity purified IgG
Safety and Immunogenicity of a Pseudomonas aeruginosa Hybrid Outer Membrane Protein F-I Vaccine in Human Volunteers
Mansouri E, et al.
Infection and Immunity, 67(7), 1461-1470 (1999)
Werner E G Müller et al.
Journal of cell science, 128(11), 2202-2207 (2015-04-25)
Polyphosphate (polyP) is a physiologically occurring polyanion that is synthesized especially in bone-forming osteoblast cells and blood platelets. We used amorphous polyP nanoparticles, complexed with Ca(2+), that have a globular size of ∼100 nm. Because polyP comprises inorganic orthophosphate units
Ana Luísa Tomás et al.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology, 39(11), 2205-2209 (2020-06-20)
Interest in the detection of specific anti-Pneumocystis jirovecii antibodies has emerged as less-invasive alternative diagnostic approaches. Here is presented the performance of an ELISA based on a recombinant synthetic multi-epitope kexin 1 (Kex1) antigen of P. jirovecii, previously developed. Results
Receptor-mediated targeting of spray-dried lipid particles coformulated with immunoglobulin and loaded with a prototype vaccine
Bot AI, et al.
Pharmaceutical Research, 18(7), 971-979 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service