Skip to Content
Merck
All Photos(1)

Documents

F5016

Sigma-Aldrich

Anti-Human IgG (Fc specific)−FITC antibody, Mouse monoclonal

clone HP-6017, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Human IgG (Fc specific)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

FITC conjugate

antibody form

purified from hybridoma cell culture

antibody product type

secondary antibodies

clone

HP-6017, monoclonal

form

buffered aqueous solution

species reactivity

rabbit, sheep, horse (IgG), goat, human

storage condition

protect from light

technique(s)

dot immunobinding: 1:16
particle immunofluorescence: 1:16

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Human IgG (mouse IgG2a isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Immunoglobulins (Igs) belongs immunoglobulin super-family. Each immunoglobin have two heavy (H) and two light (L), held together by disulphide linkages. The light comprises of one variable N-terminal region and a constant C-terminal region. The heavy chain has one variable N-terminal region and three or four constant (CH1-CH4) C-terminal region.

Specificity

The antibody is specific for the Fc portion of human IgG and recognizes an epitope common to all human IgG subclasses. This antibody was adopted as an Fc specific reagent in the IUIS/WHO study.

Application

Monoclonal Anti-Human IgG (Fc specific)-FITC antibody produced in mouse has been used in:
  • Fluorescent Dot Immunobinding Assay (F-DIBA)
  • Particle Immunofluorescent Assay (F-IFMA)
  • flow cytometry

Biochem/physiol Actions

Digestion of IgG by papain results in generation of fragment antigen binding (Fab) comprising one complete L chain and a variable and CH1 region of H chain. Pepsin digestion of IgG results in fragment crystallisable (fc), containing the H chain constant region. IgG antibody have enormous therapeutic potential and the Fc is for the development of therapeutic antibody. Although the antibody site is located in the terminal end of the human IgG molecule (part of the Fab fragment), the Fc portion has various important functions such as complement fixation, site for rheumatoid factor (autoantibodies directed to Fc), passage through placental membrane and staphylococcus protein A binding.

Physical form

Solution in 0.01 M phosphate buffer pH 8.0, containing 1% inactivated bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A nonviral carrier for targeted gene delivery to tumor cells
van Zanten J, et al.
Cancer Gene Therapy, 11(2), 156-156 (2004)
Development and characterization of clinical-grade 89Zr-trastuzumab for HER2/neu immunoPET imaging
Dijkers ECF, et al.
Journal of Nuclear Medicine, 50(6), 974-981 (2009)
Martin Pool et al.
European journal of nuclear medicine and molecular imaging, 44(8), 1328-1336 (2017-03-21)
c-MET and its ligand hepatocyte growth factor are often dysregulated in human cancers. Dynamic changes in c-MET expression occur and might predict drug efficacy or emergence of resistance. Noninvasive visualization of c-MET dynamics could therefore potentially guide c-MET-directed therapies. We
Eli C F Dijkers et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(6), 974-981 (2009-05-16)
The anti-human epidermal growth factor receptor 2 (HER2/neu) antibody trastuzumab is administered to patients with HER2/neu-overexpressing breast cancer. Whole-body noninvasive HER2/neu scintigraphy could help to assess and quantify the HER2/neu expression of all lesions, including nonaccessible metastases. The aims of
Minwoo Kang et al.
Pharmaceutics, 14(7) (2022-07-28)
Immuno-positron emission tomography (PET) has great potential to evaluate the target expression level and therapeutic response for targeted cancer therapy. Immuno-PET imaging with pertuzumab, due to specific recognition in different binding sites of HER2, could be useful for the determination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service