Skip to Content
Merck
All Photos(1)

Key Documents

A1987

Sigma-Aldrich

AM6545

≥98% (HPLC)

Synonym(s):

5-(4-[4-cyanobut-1-ynyl]phenyl)-1-(2,4-dichlorophenyl)-4-methyl-N-(1,1-dioxo-thiomorpholino)-1H-pyrazole-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H23Cl2N5O3S
CAS Number:
Molecular Weight:
556.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >25 mg/mL

storage temp.

2-8°C

SMILES string

Cc1c(nn(-c2ccc(Cl)cc2Cl)c1-c3ccc(cc3)C#CCCC#N)C(=O)NN4CCS(=O)(=O)CC4

InChI

1S/C26H23Cl2N5O3S/c1-18-24(26(34)31-32-13-15-37(35,36)16-14-32)30-33(23-11-10-21(27)17-22(23)28)25(18)20-8-6-19(7-9-20)5-3-2-4-12-29/h6-11,17H,2,4,13-16H2,1H3,(H,31,34)

InChI key

XBHQLFVDGLPBCK-UHFFFAOYSA-N

General description

AM6545 is a cannabinoid (CB1) receptor antagonist.

Biochem/physiol Actions

AM6545 helps in decreasing the development of albuminuria, inflammation and expression of markers of fibrosis. It also helps in the down-regulation of nephrin and podocin. It has the ability to repress the ingestion of food and food-reinforced behaviour.
AM6545 is a peripheral CB1 cannabinoid receptor antagonist and appetite suppressant.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Zhang et al.
Molecular medicine reports, 19(3), 1491-1500 (2018-12-21)
Increasing evidence suggests that the dysregulation of microRNAs (miRNAs) has an important role in the progression of human cancer, including ESCC. However, the exact functions and mechanisms of miRNAs in ESCC remain largely unclear. The aim of the present study
Reversal of albuminuria by combined AM6545 and perindopril therapy in experimental diabetic nephropathy
Barutta F, et al.
British Journal of Pharmacology, 175(23), 4371-4385 (2018)
The novel cannabinoid CB1 antagonist AM6545 suppresses food intake and food-reinforced behavior
Randall PA, et al.
Pharmacol. Biochem., 97(1), 179-184 (2010)
Michael Ameismeier et al.
Nature, 587(7835), 683-687 (2020-11-20)
Eukaryotic ribosomes consist of a small 40S and a large 60S subunit that are assembled in a highly coordinated manner. More than 200 factors ensure correct modification, processing and folding of ribosomal RNA and the timely incorporation of ribosomal proteins1,2.
Songqing Tang et al.
Nature communications, 5, 4657-4657 (2014-08-15)
Host immune cells can detect and destruct invading pathogens via pattern-recognition receptors. Small Rap GTPases act as conserved molecular switches coupling extracellular signals to various cellular responses, but their roles as regulators in Toll-like receptor (TLR) signalling have not been

Articles

We offer many products related to cannabinoid receptors for your research needs.

We offer many products related to cannabinoid receptors for your research needs.

We offer many products related to cannabinoid receptors for your research needs.

We offer many products related to cannabinoid receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service