Skip to Content
Merck
All Photos(1)

Documents

P57204

Sigma-Aldrich

Pyridazine

98%

Synonym(s):

1,2-Diazabenzene, 1,2-Diazine, o-Diazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2
CAS Number:
Molecular Weight:
80.09
Beilstein:
103906
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.524 (lit.)

bp

208 °C (lit.)

mp

−8 °C (lit.)

density

1.103 g/mL at 25 °C (lit.)

SMILES string

c1ccnnc1

InChI

1S/C4H4N2/c1-2-4-6-5-3-1/h1-4H

InChI key

PBMFSQRYOILNGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridazine is a mono-basic 1,2-diazine compound, which is commonly prepared by the reaction of 1,4-dicarbonyls with hydrazines. Pyridazine ring is found in many herbicides like credazine, pyridatol and many pharmaceutical drugs like cefozopran, olaparib, talazoparib, and cadralazine.

Application

Pyridazine can be used:
  • As a building block to synthesize N-phenyl-4-pyrazolo[1,5-b]pyridazin-3-yl-pyrimidin-2-amine derivatives as GSK-3 inhibitors.
  • As a starting material in the synthesis of pharmacologically important pyrrolo[1,2-b]pyridazine derivatives.
  • To prepare 1-(6-ethoxy-6-oxohexyl)pyridazin-1-ium bromide and 1-(2-bromoacetyl) pyridazinium bromide ionic liquids as corrosion inhibitors of steel under acidic conditions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

N-Phenyl-4-pyrazolo [1, 5-b] pyridazin-3-ylpyrimidin-2-amines as potent and selective inhibitors of glycogen synthase kinase 3 with good cellular efficacy
Tavares FX, et al.
Journal of Medicinal Chemistry, 47(19), 4716-4730 (2004)
Gernot Nuss et al.
Inorganic chemistry, 50(24), 12632-12640 (2011-11-19)
Reaction of potassium tris(mercapto-tert-butylpyridazinyl)borate K[Tn(tBu)] with copper(II) chloride in dichloromethane at room temperature led to the diamagnetic copper boratrane compound [Cu{B(Pn(tBu))(3)}Cl] (Pn = pyridazine-3-thionyl) (1) under activation of the B-H bond and formation of a Cu-B dative bond. In contrast
Alessio Raimondi et al.
Inorganic chemistry, 51(5), 2966-2975 (2012-03-01)
A series of [Re(2)(μ-ER)(2)(CO)(6)(μ-pydz)] complexes have been synthesized (E = S, R = C(6)H(5), 2; E = O, R = C(6)F(5), 3; C(6)H(5), 4; CH(3), and 5; H, 6), starting either from [Re(CO)(5)O(3)SCF(3)] (for 2 and 4), [Re(2)(μ-OR)(3)(CO)(6)](-) (for 3
Masaomi Terajima et al.
European journal of pharmacology, 698(1-3), 455-462 (2012-11-28)
Given the key role p38 mitogen-activated protein kinase (MAPK) plays in inflammatory responses through the production of cytokines and inflammatory mediators, its inhibition is considered a promising therapeutic strategy for chronic inflammatory diseases such as rheumatoid arthritis, psoriasis, inflammatory bowel
Scott S Walker et al.
Antimicrobial agents and chemotherapy, 55(11), 5099-5106 (2011-08-17)
The echinocandins are a class of semisynthetic natural products that target β-1,3-glucan synthase (GS). Their proven clinical efficacy combined with minimal safety issues has made the echinocandins an important asset in the management of fungal infection in a variety of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service