Skip to Content
Merck
All Photos(4)

Documents

A1260

Sigma-Aldrich

Amantadine hydrochloride

≥98% (TLC), powder, NMDA receptor antagonist

Synonym(s):

1-Adamantanamine hydrochloride, 1-Adamantylamine hydrochloride, 1-Aminoadamantane hydrochloride, NSC 83653, Tricyclo[3.3.1.13,7]decan-1-amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N · HCl
CAS Number:
Molecular Weight:
187.71
Beilstein:
4198854
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Amantadine hydrochloride,

form

powder

solubility

H2O: 50 mg/mL
ethanol: soluble

originator

Endo

SMILES string

Cl[H].[H][C@@]12C[C@@]3([H])C[C@@]([H])(C1)CC(N)(C2)C3

InChI

1S/C10H17N.ClH/c11-10-4-7-1-8(5-10)3-9(2-7)6-10;/h7-9H,1-6,11H2;1H/t7-,8+,9-,10-;

InChI key

WOLHOYHSEKDWQH-SOVZANNPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amantadine hydrochloride {tricyclo-[3,3,1,1]- deean-l-amine hydrochloride is a drug, which has rimantadine hydrochloride as its analog.

Application

Amantadine hydrochloride has been used:
  • to determine its effectiveness in reducing surgery-induced cognitive impairment
  • to preincubate multipotent stem cells (MSCs), to investigate the cellular internalization pathway
  • to determine whether amantadine-attenuated sepsis-induces neuroinflammation and dysfunction of learning and memory
  • to preincubate primary cultured rat dental pulp stem cells (rDPSCs)
  • to determine the pathway of internalization

Biochem/physiol Actions

Amantadine hydrochloride is effective against influenza viruses both in vivo and in vitro. It is considered as an antagonist of the N-methyl-D-aspartate (NMDA) type glutamate receptor. Amantadine plays an important role in the release of dopamine, preventing dopamine reuptake and blocking microglial activation and neuroinflammation.
Dopamine releaser used to treat Parkinsonism and drug-induced extrapyramidal reactions.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Endo. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Whyte et al.
Archives of physical medicine and rehabilitation, 94(10), 1877-1883 (2013-06-06)
To assess the incidence of medical complications in patients with recent traumatic disorders of consciousness (DOCs). Data on adverse events in a placebo controlled trial of amantadine hydrochloride revealed no group difference, which allowed these events to be reanalyzed descriptively
Amantadine attenuates sepsis-induced cognitive dysfunction possibly not through inhibiting toll-like receptor 2
Xing W, et al.
Journal of Molecular Medicine, 96(5), 391-402 (2018)
Intracellular co-delivery of Sr ion and phenamil drug through mesoporous bioglass nanocarriers synergizes BMP signaling and tissue mineralization
Lee J H, et al.
Acta Biomaterialia, 93-108 (2017)
Effect of aminated mesoporous bioactive glass nanoparticles on the differentiation of dental pulp stem cells
Lee J H, et al.
Testing, 11(3), e0150727-e0150727 (2016)
Guvenc Dogan et al.
Turkish neurosurgery, 30(1), 60-68 (2020-02-25)
To investigate the neuroprotective effect of a N-methyl-D-aspartate (NMDA) receptor antagonist (amantadine) in an experimental spinal cord injury (SCI) model. Thirty male Spragueâ€"Dawley rats were divided into three groups: control (I), SCI (II), and SCI + amantadine (III). SCI was

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service