Skip to Content
Merck
All Photos(1)

Key Documents

94890

Sigma-Aldrich

Victoria Blue B

for microscopy (Bact., Hist.)

Synonym(s):

Basic Blue 26, Basic Blue B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H32ClN3
CAS Number:
Molecular Weight:
506.08
Colour Index Number:
44045
Beilstein:
4094258
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.25

grade

for microscopy (Bact., Hist.)

form

powder

technique(s)

microbe id | staining: suitable

mp

206 °C (dec.) (lit.)

SMILES string

[Cl-].CN(C)c1ccc(cc1)/C(=C2/C=C\C(C=C2)=[N+](\C)C)c3ccc(Nc4ccccc4)c5ccccc35

InChI

1S/C33H31N3.ClH/c1-35(2)27-18-14-24(15-19-27)33(25-16-20-28(21-17-25)36(3)4)31-22-23-32(30-13-9-8-12-29(30)31)34-26-10-6-5-7-11-26;/h5-23H,1-4H3;1H

InChI key

LLWJPGAKXJBKKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Victoria Blue B is used as a nuclear stain in cytology/histochemical applications. Victoria Blue B may be used in the quantitative determination of phospholipids.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan-yuan Chen et al.
Guang pu xue yu guang pu fen xi = Guang pu, 26(6), 1148-1150 (2006-09-12)
In pH 4.0 acetic acid-sodium acetate buffer solution, cationic dye victory blue B shows an absorption peak at 614 nm, and the absorption peak decreases after it reacts with chondroitin sulfate to form association particles. The decrease in absorption value
Dye standards, Part II. 4: Victoria blue B (CI 44045). European Committee for Clinical Laboratory Standards (ECCLS), Subcommittee on Reference Materials for Tissue Stains (SRMTS).
The Histochemical journal, 24(4), 230-232 (1992-04-01)
Some variability factors in the cytomorphological analysis of frozen bull semen.
Galli A, Paloschi L, Pizzi F.
Andrologie, 21, 120-126 (1989)
Aikaterini Papadopoulou et al.
Regional anesthesia and pain medicine, 37(2), 183-187 (2012-02-23)
We hypothesized that by substituting a dye surrogate in place of local anesthetic, we could visually demonstrate dye sequestration by lipid emulsion that would be dependent on both dye lipophilicity and the amount of lipid emulsion used. We selected 2
S Liu et al.
Analytical chemistry, 73(16), 3907-3914 (2001-09-06)
In a near-neutral medium, a combination of heparin with some basic diphenyl naphthylmethane dyes such as victoria blue 4R (VB4R), victoria blue B (VBB), or night blue (NB) can result in a significant enhancement of resonance. Rayleigh scattering (RRS) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service