Skip to Content
Merck
All Photos(2)

Documents

73225

Supelco

Butter Yellow

analytical standard

Synonym(s):

Dimethyl yellow, 4-(Dimethylamino)azobenzene, N,N-Dimethyl-4-(phenylazo)aniline, Butter yellow, Methyl yellow

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=NC6H4N(CH3)2
CAS Number:
Molecular Weight:
225.29
Colour Index Number:
11020
Beilstein:
746016
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

111 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CN(C)c1ccc(cc1)N=Nc2ccccc2

InChI

1S/C14H15N3/c1-17(2)14-10-8-13(9-11-14)16-15-12-6-4-3-5-7-12/h3-11H,1-2H3

InChI key

JCYPECIVGRXBMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butter Yellow is a food dye widely used in industrial food production and is found to be carcinogenic in nature.

Application

Butter Yellow may be used as an analytical reference standard for the quantification of the analyte in food products using gel permeation chromatography and liquid chromatography coupled to electrospray ionization-tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gender and Well-being (2014)
NR and Proctor R
The Nazi War on Cancer (2000)
Aminoazo dye-protein-adduct enhances inhibitory effect on digestibility and damages to Gastro-Duodenal-Hepatic axis
Lin Y-L, et al.
PLoS ONE, 12(4) (2017)
Subhasish Chatterjee et al.
Nanoscale, 4(5), 1568-1571 (2012-02-03)
Self-assembly based on nucleic acid systems has become highly attractive for bottom-up fabrication of programmable matter due to the highly selective molecular recognition property of biomolecules. In this context, Y-shaped DNA (Y-DNA) provides an effective building block for forming unique
Michalina Oplatowska et al.
The Analyst, 136(11), 2403-2410 (2011-04-22)
Sensitive and specific enzyme-linked immunosorbent assays (ELISAs) were developed for the detection of two illegal synthetic dyes: Methyl Yellow (MY) and Rhodamine B (RB) in food. Polyclonal antibodies were raised against synthesised immunogens and employed in unique direct disequilibrium ELISAs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service