Skip to Content
Merck
All Photos(1)

Documents

83697

Sigma-Aldrich

Rhodamine 6G

suitable for fluorescence, BioReagent

Synonym(s):

Basic Red 1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C28H31N2O3Cl
CAS Number:
Molecular Weight:
479.01
Colour Index Number:
45160
Beilstein:
3900071
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

form

powder

solubility

H2O: soluble
ethanol: soluble

fluorescence

λex 528 nm; λem 551 nm in methanol

suitability

suitable for fluorescence

SMILES string

Cl.CCNc1cc2OC3=CC(=N/CC)\C(C)=CC3=C(c2cc1C)c4ccccc4C(=O)OCC

InChI

1S/C28H30N2O3.ClH/c1-6-29-23-15-25-21(13-17(23)4)27(19-11-9-10-12-20(19)28(31)32-8-3)22-14-18(5)24(30-7-2)16-26(22)33-25;/h9-16,29H,6-8H2,1-5H3;1H/b30-24+;

InChI key

VYXSBFYARXAAKO-WTKGSRSZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Rhodamine 6G has been used as a leukocyte marker to study the interaction between leukocytes and the vascular endothelium. It has also been used to label platelets.

Biochem/physiol Actions

Rhodamine 6G is a rhodamine analog useful in P-glycoprotein (Pgp) efflux assays. It has been used to characterize kinetics of multidrug resistance protein 1 (MRP1)-mediated efflux.
A rhodamine analog useful in Pgp efflux assays. Has been used to characterize kinetics of MRP1- mediated efflux.

Analysis Note

λmax. ∼530 nm, E(1%/1cm) ∼2000-2300 (96% Ethanol)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T P Santisakultarm et al.
Journal of thrombosis and haemostasis : JTH, 12(12), 2120-2130 (2014-09-30)
Essential thrombocythemia (ET) and polycythemia vera (PV) are myeloproliferative neoplasms (MPNs) that share the JAK2(V617F) mutation in hematopoietic stem cells, leading to excessive production of predominantly platelets in ET, and predominantly red blood cells (RBCs) in PV. The major cause
Ping Wu et al.
Analytical chemistry, 84(18), 7692-7699 (2012-08-29)
The aptamer (S2.2)-guided Ag-Au nanostructures (aptamer-Ag-Au) have been synthesized by photoreduction and validated by ultraviolet-visible light (UV-vis) spectra and transmission electron microscopy (TEM) images. Differential interference contrast (DIC), fluorescence, and TEM images, and surface-enhanced Raman scattering (SERS) spectra indicated that
MinKwan Kim et al.
Scientific reports, 5, 16525-16525 (2015-11-18)
Superresolution fluorescence microscopy possesses an important role for the study of processes in biological cells with subdiffraction resolution. Recently, superresolution methods employing the emission properties of fluorophores have rapidly evolved due to their technical simplicity and direct applicability to existing
Anasua Mukhopadhyay et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(18), 2127-2135 (2020-07-04)
Viscous, non-aqueous liquid comprising stoichiometric conjugates of polymer surfactant-bovine serum albumin (PSpBSA) is used as a host matrix for the dispersion of chemically distinct hydrophilic dyes. Using a combination of bright field polarized optical microscopy and fluorescence spectroscopy, we investigate
Asmahan Abu-Arish et al.
Biophysical journal, 97(8), 2164-2178 (2009-10-22)
The GTPase Ran is a key regulator of molecular transport through nuclear pore complex (NPC) channels. To analyze the role of Ran in its nuclear transport function, we used several quantitative fluorescence techniques to follow the distribution and dynamics of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service