Merck
  • Dispersion and Interaction of Charged Fluorescent Dyes in Protein-Polymer Surfactant-based Non-Aqueous Liquid.
Dispersion and Interaction of Charged Fluorescent Dyes in Protein-Polymer Surfactant-based Non-Aqueous Liquid.
Chemphyschem : a European journal of chemical physics and physical chemistry (2020-07-04)
Anasua Mukhopadhyay, Monika Sharma, Kamendra P Sharma
ABSTRACT
Viscous, non-aqueous liquid comprising stoichiometric conjugates of polymer surfactant-bovine serum albumin (PSpBSA) is used as a host matrix for the dispersion of chemically distinct hydrophilic dyes. Using a combination of bright field polarized optical microscopy and fluorescence spectroscopy, we investigate the dispersion of dry and powdered cationic (Rhodamine 6G; Rh6G) and anionic (Fluorescein; FL) dyes in the PSpBSA liquid at room temperature. As the dyes disperse and dissolve in the PSpBSA liquid, it results in a pronounced increase in emission intensity of the former. Interestingly, a shift from 571 to 582 nm is observed in the emission maxima of Rh6G as it disperses in the PSpBSA solvent. Whilst no such red shift is found for the Rh6G dispersion in the aqueous solutions of either native BSA or polymer-surfactant conjugated BSA, a similar shift occurs when Rh6G is dispersed in neat polymer-surfactant (PS), suggesting the interaction of the dye with the PS chains. In the case of anionic FL, no shift is observed in its emission maximum as it disperses in the PSpBSA liquid. Furthermore, within 120 minutes of FL dispersion in the PSpBSA liquid, we observe a ≈26 % decrease in the tryptophan emission intensity (λexc. =285 nm; λemi. =330 nm) of BSA, which could be attributed to both static and dynamic quenching. Our findings provide a proof of concept of an alternative non-aqueous solvent matrix which can dissolve and disperse charged fluorescent dyes, provide suitable binding sites, and show substantial photoluminescence. Thus, it can be envisaged for utilization as an alternative solvent medium for lasing dyes and related applications.
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Pricing and availability is not currently available.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Pricing and availability is not currently available.