80424
Di(N-succinimidyl) glutarate
≥97.0% (CHN)
Synonym(s):
DSG
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H14N2O8
CAS Number:
Molecular Weight:
326.26
Beilstein:
8575145
MDL number:
UNSPSC Code:
12161502
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥97.0% (CHN)
form
lumps
reaction suitability
reagent type: cross-linking reagent
storage temp.
−20°C
SMILES string
O=C(CCCC(=O)ON1C(=O)CCC1=O)ON2C(=O)CCC2=O
InChI
1S/C13H14N2O8/c16-8-4-5-9(17)14(8)22-12(20)2-1-3-13(21)23-15-10(18)6-7-11(15)19/h1-7H2
InChI key
LNQHREYHFRFJAU-UHFFFAOYSA-N
Related Categories
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Other Notes
Homobifunctional cross-linking reagent; e.g. isolation of insulin receptorcitation
Homobifunctional cross-linking reagent; e.g. isolation of insulin receptor
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
S M Waugh et al.
Biochemistry, 28(8), 3448-3455 (1989-04-18)
Radiolabeled insulin was affinity cross-linked to purified insulin receptor with six separate bifunctional N-hydroxysuccinimide esters of different lengths. Results were qualitatively identical for each cross-linker in that insulin was predominantly cross-linked through its B chain to the receptor's alpha subunit.
Jing Xu et al.
Cell discovery, 2, 16008-16008 (2016-07-28)
Mixed lineage leukemia protein-1 (MLL1) has a critical role in human MLL1 rearranged leukemia (MLLr) and is a validated therapeutic target. However, its role in regulating global gene expression in MLLr cells, as well as its interplay with MLL1 fusion
Margaret Johnson et al.
Journal of acquired immune deficiency syndromes (1999), 81(4), 463-472 (2019-04-16)
Doravirine is a novel, nonnucleoside reverse transcriptase inhibitor with demonstrated efficacy in treatment-naive adults with HIV-1. In this open-label, active-controlled, noninferiority trial, adults with HIV-1 virologically suppressed for ≥6 months on 2 nucleoside reverse transcriptase inhibitors plus a boosted protease
Anetta Ptasinska et al.
Cell reports, 28(12), 3022-3031 (2019-09-19)
Acute myeloid leukemia (AML) is associated with mutations in transcriptional and epigenetic regulator genes impairing myeloid differentiation. The t(8;21)(q22;q22) translocation generates the RUNX1-ETO fusion protein, which interferes with the hematopoietic master regulator RUNX1. We previously showed that the maintenance of
Nikki Minnebo et al.
Nucleic acids research, 41(2), 842-854 (2012-12-18)
The histone methyltransferase EZH2 regulates cell proliferation and differentiation by silencing Polycomb group target genes. NIPP1, a nuclear regulator of serine/threonine protein phosphatase 1 (PP1), has been implicated in the regulation of EZH2 occupancy at target loci, but the underlying
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service