Skip to Content
Merck
All Photos(1)

Documents

78825

Sigma-Aldrich

Liquiritigenin

≥97.0% (HPLC)

Synonym(s):

7,4′-Dihydroxyflavanone, 7-Hydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O4
Molecular Weight:
256.25
Beilstein:
359378
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.28

Assay

≥97.0% (HPLC)

form

powder or crystals

impurities

≤7% water

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

Oc1ccc(cc1)[C@@H]2CC(=O)c3ccc(O)cc3O2

InChI

1S/C15H12O4/c16-10-3-1-9(2-4-10)14-8-13(18)12-6-5-11(17)7-15(12)19-14/h1-7,14,16-17H,8H2/t14-/m0/s1

InChI key

FURUXTVZLHCCNA-AWEZNQCLSA-N

General description

Liquiritigenin is a flavonoid and an estrogenic compound found in licorice (Glycyrrhizae radix) root extract and several other plants.

Application

Liquiritigenin has been used:
  • to study its inhibitory effect on tumor metastasis in the treatment of colorectal cancer
  • as a reference standard for ultra-performance liquid chromatography (UPLC) of Chaihu-Shugan-San (CSS) extract
  • as a potential antiviral drug against hepatitis C virus (HCV) infection

Biochem/physiol Actions

Liquiritigenin displays anti-diabetic and choleretic properties. It exerts anti-inflammatory activity on Raw246.7 cells by inhibiting nuclear factor kappa light chain enhancer of activated B cells (NF-κB)-dependent-induction of inducible NOS (iNOS). Liquiritigenin inhibits liver fibrogenesis by blocking Hippo/Yes-associated protein (YAP) and transforming growth factor-β1 (TGF-β1)/small mothers against decapentaplegic (Smad) components. It is a selective estrogen receptor β agonist cells. Liquiritigenin induces apoptosis in SMM-721 cells by disruption of the mitochondrial membrane potential and increased production of reactive oxygen species.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Casey L Sayre et al.
Biomedical chromatography : BMC, 27(3), 404-406 (2012-07-21)
Pharmacometric characterization studies of liquiritigenin have historically overlooked its chiral nature. To achieve complete characterization, an analytical method enabling the detection and quantification of the individual enantiomers of racemic (±) liquiritigenin is necessary. Resolution of the enantiomers of liquiritigenin was
Rui Ting Liu et al.
European journal of pharmacology, 669(1-3), 76-83 (2011-08-30)
The present paper is to examine whether liquiritigenin is able to attenuate the Alzheimer's-like learning and memory deficits in a transgenic (Tg) mouse model that over-expresses amyloid protein precursor (APP), and explores the underlying mechanisms. Consistent with our previous observations
Suengmok Cho et al.
Bioorganic & medicinal chemistry, 20(11), 3493-3501 (2012-05-01)
Licorice (Glycyrrhiza glabra, GG) is one of the most frequently used herbal medicines worldwide, and its various biological activities have been widely studied. GG is reported to have neurological properties such as antidepressant, anxiolytic, and anticonvulsant effects. However, its hypnotic
Jennifer E Mersereau et al.
Molecular and cellular endocrinology, 283(1-2), 49-57 (2008-01-08)
After the Women's Health Initiative found that the risks of hormone therapy outweighed the benefits, a need for alternative drugs to treat menopausal symptoms has emerged. We explored the possibility that botanical agents used in Traditional Chinese Medicine for menopausal
Eun Mi Choi
International immunopharmacology, 12(1), 139-143 (2011-11-26)
Liquiritigenin is one of the flavonoids present in Glycyrrhizae radix. In the present study, the effects of liquiritigenin on the function of osteoblastic MC3T3-E1 cells were studied. Liquiritigenin caused a significant elevation of cell growth, alkaline phosphatase activity, collagen synthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service