Skip to Content
Merck
All Photos(1)

Key Documents

78544

Sigma-Aldrich

Glucose tetrasaccharide

≥99.0% (TLC)

Synonym(s):

Glcα1-6Glcα1-4Glcα1-4Glc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H42O21
CAS Number:
Molecular Weight:
666.58
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.28

Assay

≥99.0% (TLC)

form

powder

color

white

suitability

corresponds for LC-MS

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](OC[C@H]2O[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)C(O)O[C@@H]4CO)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C24H42O21/c25-1-5-9(28)11(30)16(35)22(41-5)39-4-8-10(29)12(31)17(36)23(43-8)45-20-7(3-27)42-24(18(37)14(20)33)44-19-6(2-26)40-21(38)15(34)13(19)32/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10-,11+,12+,13-,14-,15-,16-,17-,18-,19-,20-,21?,22+,23-,24-/m1/s1

InChI key

FPBCRLIOSBQLHS-QVTSYAGHSA-N

Application

Glucose tetrasaccharide (Glc4) is used as a reference in analysis of urinary glucose tetrasacchaide, a biomarker for Pompe disease and other glycogen storage diseases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chihiro Yonee et al.
Brain & development, 34(10), 834-839 (2012-04-24)
Pompe disease is an autosomal recessive disorder caused by deficiency of the lysosomal enzyme, acid alpha-glucosidase (GAA). To the best of our knowledge, no studies have reported the results of systematic and sequential CT analyses before and during ERT. In
D A Zopf et al.
Journal of immunological methods, 48(1), 109-119 (1982-01-01)
A radioimmunoassay is described that allows rapid determination of a urinary oligosaccharide -- Glc alpha 1-6Glc alpha 1-4Glc alpha 1-4Glc [(Glc)4] -- at concentrations greater than 2 pmol/microliter. Antibodies produced in rabbits immunized with the phenethylamine derivative of (Glc)4 coupled
Unique oligosaccharide (apparently glucotetrasaccharide) in urine of patients with glycogen storage diseases.
K Oberholzer et al.
Clinical chemistry, 36(7), 1381-1381 (1990-07-01)
A Lundblad et al.
Journal of immunological methods, 68(1-2), 217-226 (1984-03-30)
Monoclonal antibodies were produced in BALB/cJ mice against a urinary glucose-containing tetrasaccharide (Glc)4 coupled to keyhole limpet hemocyanin (KLH). The immune response was studied using 7 different strains of mice and 11 different immunization protocols. From 6 fusions using the
Mikael Schwarz et al.
Journal of the neurological sciences, 244(1-2), 59-68 (2006-02-17)
There is an unmet need to develop specific biomarkers for multiple sclerosis (MS) to aid in the diagnosis, improve the management of patients and the monitoring of the effectiveness of treatment. We have screened serum from patients with relapsing-remitting MS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service