Skip to Content
Merck
All Photos(1)

Documents

71515

Sigma-Aldrich

Sodium pyrophosphate tetrabasic decahydrate

BioUltra, ≥99.5% (T)

Synonym(s):

Pyrophosphate inorganic, Sodium diphosphate tetrabasic, Tetrasodium pyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na4P2O7 · 10H2O
CAS Number:
Molecular Weight:
446.06
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.21

product line

BioUltra

Assay

≥99.5% (T)

form

crystals

impurities

insoluble matter, passes filter test

pH

9.5-11.0 (25 °C, 0.1 M in H2O)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.010
λ: 280 nm Amax: 0.010

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/4Na.H4O7P2.10H2O/c;;;;1-8(2,3)7-9(4,5)6;;;;;;;;;;/h;;;;(H2,1,2,3)(H2,4,5,6);10*1H2/q4*+1;;;;;;;;;;;/p-4

InChI key

VZWGHDYJGOMEKT-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Other Notes

In assays involving ATP-pyrophosphate exchange - tryptophanyl-tRNA synthesis; Enzyme assay in which the product pyrophosphate is measured, addition of carrier pyrophosphate insures quantitative precipitation

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tryptophanyl-tRNA synthetase from beef pancreas.
L L Kisselev et al.
Methods in enzymology, 59, 234-257 (1979-01-01)
Amidophosphoribosyltransferase (chicken liver).
J M Lewis et al.
Methods in enzymology, 51, 171-178 (1978-01-01)
Taís Scaramucci et al.
Journal of dentistry, 39(12), 841-848 (2011-09-29)
To evaluate the erosive potential of orange juice modified by food additives in enamel and dentine. Calcium lactate pentahydrate (CLP), xanthan gum (XG), sodium linear polyphosphate (LPP), sodium pyrophosphate tetrabasic (PP), sodium tripolyphosphate (STP) and some of their combinations were
Priyadip Das et al.
Chemical communications (Cambridge, England), 47(28), 8118-8120 (2011-06-21)
Newly synthesized Zn(II) and Cd(II)-based complexes show unique selectivity towards inorganic pyrophosphate (PPi) in 100% aqueous medium at pH = 7.4 and act as a "turn-on" and "turn-off" real-time assay, respectively, for the evaluation of the enzymatic activity of alkaline
Nadia Marino et al.
Inorganic chemistry, 50(1), 378-389 (2010-12-01)
The reaction in water of Cu(NO(3))(2)·2.5H(2)O with 2,2'-bipyridine (bipy), 1,10-phenanthroline (phen), or 1,10-phenanthroline-5-amine (phenam), and sodium pyrophosphate (Na(4)P(2)O(7)), at various pHs, afforded three new copper(II)-pyrophosphate complexes, namely, {[Cu(bipy)(cis-H(2)P(2)O(7))](2)}·3H(2)O (1a), {[Cu(phen)(H(2)O)](4)(HP(2)O(7))(2)}(ClO(4))(2)·4H(2)O (2), and {[Cu(2)(phenam)(2)(P(2)O(7))](2)·25H(2)O}(n) (3). A solvent free crystalline phase of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service