Skip to Content
Merck
All Photos(1)

Documents

71501

Sigma-Aldrich

Sodium pyrophosphate dibasic

BioUltra, ≥99.0% (T)

Synonym(s):

Disodium dihydrogen pyrophosphate, Sodium diphosphate dibasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2H2P2O7
CAS Number:
Molecular Weight:
221.94
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥99.0% (T)

form

powder

impurities

insoluble matter, passes filter test

loss

≤0.5% loss on drying, 200 °C

pH

3.5-4.5 (20 °C, 0.1 M in H2O, freshly prepared)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.2 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤300 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.11
λ: 280 nm Amax: 0.09

SMILES string

[Na+].[Na+].OP([O-])(=O)OP(O)([O-])=O

InChI

1S/2Na.H4O7P2/c;;1-8(2,3)7-9(4,5)6/h;;(H2,1,2,3)(H2,4,5,6)/q2*+1;/p-2

InChI key

GYQBBRRVRKFJRG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium pyrophosphate dibasic has been used:
  • as a setting retardant
  • as a component of STOP-washing buffer for in gel kinase assay
  • as protease and phosphatase inhibitor in the preparation of western blot lysis buffer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin tolerance test under anaesthesia to measure tissue-specific insulin-stimulated glucose disposal
Fazakerley DJ, et al.
eLife (2018)
Brushite foams?the effect of T ween
Unosson J, et al.
Journal of biomedical research, 104(1), 67-77 (2016)
In Gel Kinase Assay
Pizzio GA and Rodriguez PL
Bio-protocol (2017)
Caroline Öhman et al.
Journal of materials science. Materials in medicine, 26(7), 210-210 (2015-07-15)
The porosity of calcium phosphate cements has an impact on several important parameters, such as strength, resorbability and bioactivity. A model to predict the porosity for biomedical cements would hence be a useful tool. At the moment such a model
Jonathan A Stefely et al.
Autophagy, 16(12), 2219-2237 (2020-01-24)
Macroautophagy/autophagy is suppressed by MTOR (mechanistic target of rapamycin kinase) and is an anticancer target under active investigation. Yet, MTOR-regulated autophagy remains incompletely mapped. We used proteomic profiling to identify proteins in the MTOR-autophagy axis. Wild-type (WT) mouse cell lines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service