Skip to Content
Merck
All Photos(1)

Documents

66960

Sigma-Aldrich

4-O-Methyl-D-glucurono-D-xylan dyed with Remazol brilliant blue R

endo-l,4-β-xylanase substrate, fibers

Synonym(s):

RBB-Xylan, Remazol brilliant blue R–D-Xylan

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

product name

4-O-Methyl-D-glucurono-D-xylan dyed with Remazol brilliant blue R, 15% dye basis

form

fibers

concentration

15% (dye)

solubility

water: 0.05 g/5mL, blue to very deep blue

Application

4-o-Methyl-D-glucurono-D-xylan dyed with Remazol brilliant blue R has been used as a substrate for endo-l,4-β-xylanase activity.

Other Notes

Soluble chromogenic substrate for the assay of endo-1,4-β-xylanases

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermostable xylanolytic enzymes from Rhodothermus marinus grown on xylan
Dahlberg L, et al.
Applied Microbiology and Biotechnology, 40(1), 63-68 (1993)
P. Biely et al.
Methods in Enzymology, 160, 536-536 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service