D133000
3,4-Dimethoxybenzyl alcohol
96%
Synonym(s):
(3,4-Dimethoxyphenyl)methanol, Veratryl alcohol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(CH3O)2C6H3CH2OH
CAS Number:
Molecular Weight:
168.19
Beilstein:
639388
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
96%
form
liquid
refractive index
n20/D 1.552 (lit.)
bp
296-297 °C/732 mmHg (lit.)
density
1.157 g/mL at 25 °C (lit.)
SMILES string
COc1ccc(CO)cc1OC
InChI
1S/C9H12O3/c1-11-8-4-3-7(6-10)5-9(8)12-2/h3-5,10H,6H2,1-2H3
InChI key
OEGPRYNGFWGMMV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
3,4-Dimethoxybenzyl alcohol is widely used in the synthesis of various cyclotriveratrylenes (CTVs), which are cyclic molecular hosts with a cavity to accommodate guest molecules. It can also be used as a precursor in the total synthesis of salvianolic acid N.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Recent advances in the chemistry of cyclotriveratrylene.
Hardie M J
Chemical Society Reviews, 39(2), 516-527 (2010)
Self-assembly of dendritic crowns into chiral supramolecular spheres.
Percec V, et al.
Journal of the American Chemical Society, 131(3), 1294-1304 (2008)
Improved synthesis of functional CTVs and cryptophanes using Sc (OTf) 3 as catalyst.
Brotin T, et al.
The Journal of Organic Chemistry, 70(16), 6187-6195 (2005)
Formal total synthesis of salvianolic acid N.
Wu K, et al.
Organic & Biomolecular Chemistry (2018)
Yongjun Zhang et al.
Journal of hazardous materials, 176(1-3), 1089-1092 (2009-12-01)
Carbamazepine and diclofenac were frequently detected in water bodies. In this study, crude lignin peroxidase, produced from a white rot fungus Phanerochaete chrysosporium, was studied on its in vitro degradation of both drugs. The influencing parameters were studied, including pH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service