Skip to Content
Merck
All Photos(1)

Documents

65906

Sigma-Aldrich

Phalloidin–Atto 647N

BioReagent, suitable for fluorescence, ≥80% (HPLC)

Synonym(s):

Atto 647N, Atto 647N-Phalloidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171501
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥80% (HPLC)

manufacturer/tradename

ATTO-TEC GmbH

λ

in methanol

UV absorption

λ: 640-646 nm Amax

suitability

suitable for fluorescence

detection method

fluorometric

storage temp.

−20°C

General description

Phalloidin–Atto 647N is a new fluorescentlabel targeting the red spectral region. Atto 647N is a cationic dye, and postcoupling, it carries a net electrical charge of +1.Like most Atto labels, the absorption andfluorescence of Atto 647N are independent of pH between 2-11. Atto 647N issupplied in the form of a mixture containing two isomers having identicalfluorescence and absorption properties. Atto labels have rigid structures thatdo not show any cis-trans-isomerization.

Application

Phalloidin–Atto 647Nis designed to be used for labelling DNA, RNA, or proteins. Fluorescentconjugates of phalloidin are used to label actin filaments for histologicalapplications. Some structural features of phalloidin are required for thebinding to actin.

Features and Benefits

Characteristic features of the Phalloidin Atto488 are:
  1. StrongAbsorption.
  2. HighFluorescence quantum yield.
  3. HighPhotostability.
  4. MinimalTriplet formation.
  5. GoodSolubility.
  6. Excellent Ozone Resistance.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georgios Trichas et al.
BMC biology, 6, 40-40 (2008-09-17)
Transgenic animals are widely used in biomedical research and biotechnology. Multicistronic constructs, in which several proteins are encoded by a single messenger RNA, are commonly used in genetically engineered animals. This is currently done by using an internal ribosomal entry
Catherine Pfefferli et al.
Nature communications, 8, 15151-15151 (2017-05-04)
The existence of common mechanisms regulating organ regeneration is an intriguing concept. Here we report on a regulatory element that is transiently activated during heart and fin regeneration in zebrafish. This element contains a ctgfa upstream sequence, called careg, which
Benedetta Artegiani et al.
Nature cell biology, 22(3), 321-331 (2020-03-04)
CRISPR-Cas9 technology has revolutionized genome editing and is applicable to the organoid field. However, precise integration of exogenous DNA sequences into human organoids is lacking robust knock-in approaches. Here, we describe CRISPR-Cas9-mediated homology-independent organoid transgenesis (CRISPR-HOT), which enables efficient generation
Francesca Farina et al.
The EMBO journal, 38(11) (2019-04-25)
Cells going through mitosis undergo precisely timed changes in cell shape and organisation, which serve to ensure the fair partitioning of cellular components into the two daughter cells. These structural changes are driven by changes in actin filament and microtubule
Maria-Del-Carmen Diaz-de-la-Loza et al.
Development (Cambridge, England), 147(5) (2020-03-04)
Mutations in the Ultrabithorax (Ubx) gene cause homeotic transformation of the normally two-winged Drosophila into a four-winged mutant fly. Ubx encodes a HOX family transcription factor that specifies segment identity, including transformation of the second set of wings into rudimentary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service