Skip to Content
Merck
All Photos(3)

Documents

60339

Sigma-Aldrich

Potassium bicarbonate

BioUltra, ≥99.5% (T)

Synonym(s):

Potassium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHCO3
CAS Number:
Molecular Weight:
100.12
Beilstein:
4535309
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Assay

≥99.5% (T)

form

powder or crystals

impurities

insoluble matter, passes filter test

pH

7.5-9.0 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.010
λ: 280 nm Amax: 0.01

SMILES string

[K+].OC([O-])=O

InChI

1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

TYJJADVDDVDEDZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium bicarbonate may be used with D-ribose to induce a synergistic effect on the slowing of cancer cell line proliferation in vitro. Potassium bicarbonate is used in studies on the effect of bicarbonate salts on protein ligand binding.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amalie Modvig et al.
Materials (Basel, Switzerland), 12(13) (2019-07-10)
A Ru-doped phospho-tungstic Wells-Dawson polyoxometalate (POM) was successfully applied as homogeneous catalyst for glycerol hydrogenolysis in aqueous media. The synthesized compound showed superior catalytic activity compared to classical homogeneous/heterogeneous Ru catalysts like RuCl3 and Ru/C under identical reaction conditions, whereas
Francesco Orsini et al.
Methods (San Diego, Calif.), 51(1), 106-113 (2009-12-10)
This paper describes the use of Atomic Force Microscopy (AFM) to investigate the plasma membrane of Xenopus laevis oocyte. Different protocols of sample preparation to perform an AFM investigation of both external and intracellular sides of the oocyte plasma membrane
Daniel L Belavý et al.
Journal of applied physiology (Bethesda, Md. : 1985), 111(5), 1304-1314 (2011-07-30)
As part of the nutrition-countermeasures (NUC) study in Cologne, Germany in 2010, seven healthy male subjects underwent 21 days of head-down tilt bed rest and returned 153 days later to undergo a second bout of 21-day bed rest. As part
Lisa Ceglia et al.
The Journal of clinical endocrinology and metabolism, 94(2), 645-653 (2008-12-04)
Protein is an essential component of muscle and bone. However, the acidic byproducts of protein metabolism may have a negative impact on the musculoskeletal system, particularly in older individuals with declining renal function. We sought to determine whether adding an
Masaru Watanabe et al.
Carbohydrate research, 341(18), 2891-2900 (2006-11-01)
Liquefaction of glucose into oil was examined in hot-compressed water at 300 degrees C and 30 or 60 min in a tumbling batch reactor. The effects of alkali (KHCO(3)), a hydrogenating agent (HCO(2)H), and a cobalt catalyst (Co(3)O(4)) were studied.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service