Skip to Content
Merck
All Photos(1)

Key Documents

60010

Sigma-Aldrich

Kaempferol

≥97.0% (HPLC)

Synonym(s):

3,4′,5,7-Tetrahydroxyflavone, 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Robigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O6
CAS Number:
Molecular Weight:
286.24
Beilstein:
304401
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.47

Assay

≥97.0% (HPLC)

impurities

≤12% water

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

Oc1ccc(cc1)C2=C(O)C(=O)c3c(O)cc(O)cc3O2

InChI

1S/C15H10O6/c16-8-3-1-7(2-4-8)15-14(20)13(19)12-10(18)5-9(17)6-11(12)21-15/h1-6,16-18,20H

InChI key

IYRMWMYZSQPJKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kaempferol is a polyphenolic antioxidant abundantly present in vegetables and fruits. It has a diphenylpropane structure. In many plants, it is present as a glycosidic form namely, kaempferol-3-O-glucoside.

Application

Chromogenic reagent for antimony in the low ppm range and for gallium and indium in the sub-ppm range.
Kaempferol has been used:
  • to test its effect on osteoblast proliferation, migration, and differentiation in mouse pre-osteoblast cell line (MC3T3-E1 cells)
  • as an antioxidant and a hepatoprotective agent against zearalenone (ZEA)-induced oxidative stress and apoptosis in HEPG2 cells
  • as a neuroprotective agent against cadmium chloride (CdCl2) induced hippocampal damage and memory deficits in rats
  • as an anti-inflammatory agent, together with apigenin, for stem-cell therapy on knee osteoarthritic male rats

Biochem/physiol Actions

Kaempferol is hydrophobic in nature due to its diphenylpropane group. It exerts anti-inflammatory actions along with their antioxidant properties and modulates pro-inflammatory enzyme activities. Kaempferol acts as a chemopreventive agent by exerting protective effects on cell viability. It modulates several key elements of cellular signal transduction pathways linked to apoptosis, angiogenesis, metastasis, and inflammation.
Potent inhibitor of osteoclastic bone resorption. The effect is believed to be attributable to both the antioxidant and estrogenic activities of kaempferol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Firoozeh Estakhri et al.
The Knee, 27(3), 817-832 (2020-04-28)
Based on the anti-inflammatory and anti-oxidant properties of kaempferol and apigenin, we hypothesized that co-injection of these phytochemicals would increase the effectiveness of cell therapy in knee osteoarthritic rats. Anterior cruciate ligament transection (ACLT) was used to induce osteoarthritis (OA).
Kaempferol protects against cadmium chloride-induced hippocampal damage and memory deficits by activation of silent information regulator 1 and inhibition of poly (ADP-Ribose) polymerase-1
El-kott A F, et al.,
The Science of the Total Environment, 138832-138832 (2020)
Kasi Pandima Devi et al.
Pharmacological research, 99, 1-10 (2015-05-20)
Inflammation is an important process of human healing response, wherein the tissues respond to injuries induced by many agents including pathogens. It is characterized by pain, redness and heat in the injured tissues. Chronic inflammation seems to be associated with
Yang Wang et al.
Artificial cells, nanomedicine, and biotechnology, 47(1), 1050-1056 (2019-04-04)
Osteoblast has been found to exert indispensable functions during fracture healing. This study aimed to investigate the effects of kaempferol, a natural flavonoid compound, on osteoblast proliferation, migration and differentiation, as well as possible molecular mechanisms. qRT-PCR was performed to
Allen Y Chen et al.
Food chemistry, 138(4), 2099-2107 (2013-03-19)
Kaempferol is a polyphenol antioxidant found in fruits and vegetables. Many studies have described the beneficial effects of dietary kaempferol in reducing the risk of chronic diseases, especially cancer. Epidemiological studies have shown an inverse relationship between kaempferol intake and

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service