Skip to Content
Merck
All Photos(1)

Key Documents

420345

Sigma-Aldrich

Kaempferol

A cell-permeable phytoestrogen that inhibits topoisomerase I-catalyzed DNA religation in HL-60 cells.

Synonym(s):

Kaempferol, 3,4ʹ,5,7-Tetrahydroxyflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O6
CAS Number:
Molecular Weight:
286.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 5274

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

ethanol: 20 mg/mL
DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[o]1c2c([c](c(c1c3ccc(cc3)O)O)=O)c(cc(c2)O)O

InChI

1S/C15H10O6/c16-8-3-1-7(2-4-8)15-14(20)13(19)12-10(18)5-9(17)6-11(12)21-15/h1-6,16-18,20H

InChI key

IYRMWMYZSQPJKC-UHFFFAOYSA-N

General description

A cell-permeable phytoestrogen that inhibits topoisomerase I-catalyzed DNA religation in HL-60 cells. Offers protection against Aβ25-35-induced cell death in neonatal cortical neurons. Its protective effects are comparable to that of estradiol. Blocks the Aβ-induced activation of caspase-2, -3, -8, and -9, and reduces NMDA-induced neuronal apoptosis. Reported to be a potent inhibitor of monoamine oxidases. Acts as an inhibitor of COX-1 activity (IC50 = 180 µM), and of transcriptional activation of COX-2 (IC50<15 µM).
A cell-permeable, phytoestrogen that offers protection against Aβ25-35-induced cell death in neonatal cortical neurons. Its protective effects are comparable to that of estradiol. Reduces the activation of caspases 2, 3, 8, and 9 by Aβ. Inhibits topoisomerase I-catalyzed DNA religation in HL-60 cells. Also shown to protect against NMDA-induced neuronal toxicity in vitro in cultured rat cortical neurons. Acts as an inhibitor of cyclooxygenases (COX-1, IC50 = 180 µM) and COX-2 (IC50 = ~15 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Topoisomerase I-catalyzed DNA religation in HL-60 cells
Product does not compete with ATP.
Reversible: no
Target IC50: 180 µM against COX-1 activity

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wang, C.N., et al. 2001. J. Biol. Chem.276, 5287.
Sloley, B.D., et al. 2000. J. Pharm. Pharmacol.52, 451.
Wang, H., et al. 2000. Phytomedicine7, 15.
Liang, Y.C., et al. 1999. Carcinogenesis20, 1945.
Roth, A., et al. 1999. J. Neurosci. Res.57, 399.
Boege, F., et al. 1996. J. Biol. Chem.271, 2262.
Constantinou, A., et al. 1995. J. Nat. Prod.58, 217.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service