Skip to Content
Merck
All Photos(1)

Key Documents

54274

Sigma-Aldrich

Chelidonine

≥97.0% (HPLC)

Synonym(s):

Stylophorin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19NO5
CAS Number:
Molecular Weight:
353.37
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97.0% (HPLC)

form

solid

impurities

~5% water

storage temp.

2-8°C

SMILES string

[H][C@]12[C@@H](O)Cc3cc4OCOc4cc3[C@@]1([H])N(C)Cc5c6OCOc6ccc25

InChI

1S/C20H19NO5/c1-21-7-13-11(2-3-15-20(13)26-9-23-15)18-14(22)4-10-5-16-17(25-8-24-16)6-12(10)19(18)21/h2-3,5-6,14,18-19,22H,4,7-9H2,1H3/t14-,18-,19+/m0/s1

InChI key

GHKISGDRQRSCII-ZOCIIQOWSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits tubulin polymerisation (IC50=24 μM), thereby disrupting microtubule structure in cells and inducing a G2/M mitotic arrest.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Grynkiewicz et al.
European journal of medicinal chemistry, 36(11-12), 951-960 (2002-01-05)
A group of 11 derivatives of chelidonine was obtained by acylations and/or alkylations of the secondary hydroxyl group with the aim of testing their biological activity. This paper focuses on the new derivatives influence on CNS in mice. The highest
Alex Philchenkov et al.
Toxicology in vitro : an international journal published in association with BIBRA, 22(2), 287-295 (2007-11-21)
Apoptogenic and DNA damaging effects of chelidonine (CHE) and sanguinarine (SAN), two structurally related benzophenanthridine alkaloids isolated from Chelidonium majus L. (Papaveraceae), were compared. Both alkaloids induced apoptosis in human acute T-lymphoblastic leukaemia MT-4 cells. Apoptosis induction by CHE and
Mahmoud Zaki El-Readi et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 20(3-4), 282-294 (2012-12-15)
Cancer cells often develop multidrug resistance (MDR) which is a multidimensional problem involving several mechanisms and targets. This study demonstrates that chelidonine and an alkaloid extract from Chelidonium majus, which contains protoberberine and benzo[c]phenanthridine alkaloids, has the ability to overcome
J Wolff et al.
Biochemistry, 32(48), 13334-13339 (1993-12-07)
Chelidonine, sanguinarine, and chelerythrine are natural benzophenanthridine alkaloids that inhibit taxol-mediated polymerization of rat brain tubulin in the micromolar range. Chelidonine is a weak, competitive inhibitor of colchicine binding to tubulin but does not inhibit podophyllotoxin binding. On the other
Matthew J Fleming et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(7), 2112-2124 (2008-01-18)
New enantioselective syntheses of the B/C hexahydrobenzo[c]phenanthridine alkaloids (+)-homochelidonine, (+)-chelamidine, (+)-chelidonine, (+)-chelamine, and (+)-norchelidonine are described. Our rapid and convergent route to this class of natural products involved the development and application of a Pd II-catalyzed asymmetric ring-opening reaction of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service