Skip to Content
Merck
All Photos(1)

Key Documents

SMB00377

Sigma-Aldrich

Magnoflorine

≥98% (HPLC)

Synonym(s):

(6aS)-2,10-dimethoxy-6,6-dimethyl-5,6,6a,7-tetrahydro-4H-dibenzo[de,g]quinoline-6-ium-1,11-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24NO4
CAS Number:
Molecular Weight:
342.41
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

OC1=C(OC)C=C2CC[N+](C)(C)[C@@]3([H])CC4=CC=C(OC)C(O)=C4C1=C32

InChI

1S/C20H23NO4/c1-21(2)8-7-12-10-15(25-4)20(23)18-16(12)13(21)9-11-5-6-14(24-3)19(22)17(11)18/h5-6,10,13H,7-9H2,1-4H3,(H-,22,23)/p+1/t13-/m0/s1

InChI key

YLRXAIKMLINXQY-ZDUSSCGKSA-O

General description

Magnoflorine is a secondary metabolite and an alkaloid with a quaternary aporphine configuration. It can be found in roots, rhizomes, stems, barks, or seeds of many herbal plants such as Caulophyllum thalictroides (blue cohosh), Arisolochai bracteate, P. amurense, S. acutum, Tinospora crispa, Coptidis rhizome, etc. Magnoflorine constitutes approximately 65 % of the total alkaloids in roots and rhizomes of blue cohosh. It is also known as thalictrine and escholine.

Application

Magnoflorin has been used to test its contractile effects on isolated mouse uterine smooth muscle tissues and on isolated strips of Mus musculus distal colon smooth muscle tissues.

Biochem/physiol Actions

Magnoflorine possesses several biochemical and pharmacological properties such as antioxidant, anti-inflammatory, immunomodulatory, anti-diabetic, anti-fungal, cardiovascular protective, and neuropsychological. It also acts as an α-tyrosinase inhibitor. It helps to increase pro-inflammatory responses induced by lipopolysaccharide (LPS). It exerts antitumor, anti-cancer activities by inhibiting gastric cancer progression gastric cancer cell xenograft mouse model.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tong Xu et al.
Pharmacological research, 152, 104632-104632 (2020-01-09)
Magnoflorine is an important quaternary aporphine alkaloid that is isolated from some commonly used herbal medicines (e.g., Sinomenium acutum (Thunb.) Rehder & E.H.Wilson and Coptis chinensis Franch.). In recent years, magnoflorine has received increasing attention due to its multiple pharmacological
Quillaja saponins are a potent contractor of uterine smooth muscle tissue in vitro
Bristol, Brian and Degolier, Teresa
Journal of Pharmacognosy and Phytochemistry, 7(5), 1252-1258 (2018)
Potential biological activities of magnoflorine: a compound from Aristolochia debilis Sieb. et Zucc
Li C and Wang MH
The Korean Journal of Plant Resources, 27(3), 223-228 (2014)
Md Areeful Haque et al.
Planta medica, 84(17), 1255-1264 (2018-06-16)
Magnoflorine, a major bioactive metabolite isolated from Tinospora crispa, has been reported for its diverse biochemical and pharmacological properties. However, there is little report on its underlying mechanisms of action on immune responses, particularly on macrophage activation. In this study
The contractile responses of blue cohosh and some its constituents on isolated longitudinal strips of mouse distal colon
Cermin, Hayley and DeGolier, Teresa
Journal of Pharmacognosy and Phytochemistry, 8(2), 2160-2166 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service