Skip to Content
Merck
All Photos(1)

Documents

39693

Sigma-Aldrich

(±)-Sodium 2,3-dihydroxyisovalerate hydrate

≥95% (CE)

Synonym(s):

Sodium 2,3-dihydroxy-3-methylbutanoate hydrate, Sodium DL-α,β-dihydroxyisovalerate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9O4Na · xH2O
CAS Number:
Molecular Weight:
156.11 (anhydrous basis)
Beilstein:
1722373
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (CE)

form

powder or crystals

impurities

~12% water

storage temp.

2-8°C

SMILES string

O.[Na+].CC(C)(O)C(O)C([O-])=O

InChI

1S/C5H10O4.Na.H2O/c1-5(2,9)3(6)4(7)8;;/h3,6,9H,1-2H3,(H,7,8);;1H2/q;+1;/p-1

InChI key

KGRRZNPTTZIUMS-UHFFFAOYSA-M

Biochem/physiol Actions

Racemic form of intermediate in L-valine catabolism and anabolism.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence for an enol intermediate in the enzymatic conversion of alpha,beta-dihydroxyisovalerate to alpha-ketoisovalerate.
S M Arfin
The Journal of biological chemistry, 244(9), 2250-2251 (1969-05-10)
F M van der Kloet et al.
Metabolomics : Official journal of the Metabolomic Society, 8(1), 109-119 (2012-01-27)
Diabetic kidney disease (DKD) is a devastating complication that affects an estimated third of patients with type 1 diabetes mellitus (DM). There is no cure once the disease is diagnosed, but early treatment at a sub-clinical stage can prevent or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service