Skip to Content
Merck
All Photos(2)

Key Documents

390A

Sigma-Aldrich

Leukocyte Peroxidase (Myeloperoxidase) Kit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352106
NACRES:
NA.47

description

Hanker-Yates substrate kit

Quality Level

usage

sufficient for 10-120 manual assays

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

The Leukocyte Peroxidase (Myeloperoxidase) kit is intended for use as a general laboratory histochemical stain for the demonstration of leukocyte peroxidase. This manual, qualitative procedure identifies leukocyte peroxidase in blood or bone marrow films of human specimens.

Kit Components Only

Product No.
Description

  • Acid Hematoxylin Solution (2852) 100 mL

  • Peroxidase Indicator Reagent (3901) 10 vial(s)

Kit Components Also Available Separately

Product No.
Description
SDS

  • 903CTRIZMAL™ 6.3 Buffer Concentrate, pH 6.3(concentrate), 0.2 M 50 mLSDS

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natan Krohn et al.
Gastroenterology, 136(5), 1806-1817 (2009-05-08)
Hepatocyte transplantation-induced liver inflammation impairs cell engraftment. We defined whether proinflammatory cytokines and chemokines played roles in regulation of hepatocyte engraftment in the liver. We performed studies over up to 3 weeks in rat hepatocyte transplantation systems. Expression of 84
Rui Zhang et al.
Cell death & disease, 11(4), 277-277 (2020-04-26)
DNA damage regulated autophagy modulator 1 (DRAM1) is a stress-inducible regulator of autophagy and cell death. DRAM1 has been implicated in cancer, myocardial infarction, and infectious diseases, but the molecular and cellular functions of this transmembrane protein remain poorly understood.
Emily A Beck et al.
G3 (Bethesda, Md.), 10(2), 613-622 (2019-12-18)
Selection, via host immunity, is often required to foster beneficial microbial symbionts and suppress deleterious pathogens. In animals, the host immune system is at the center of this relationship. Failed host immune system-microbial interactions can result in a persistent inflammatory
Tracy Stokol et al.
Journal of the American Association for Laboratory Animal Science : JAALAS, 60(6), 616-629 (2021-08-29)
Southern giant pouched rats (Cricetomys ansorgei) are muroid rodents native to subSaharan Africa. They are increasingly used as service animals because of their keen sense of smell and are primarily known for clearing minefields in Africa. The objectives of this
Yuta Enami et al.
Gastroenterology, 136(7), 2356-2364 (2009-03-24)
Hepatic inflammation occurs immediately after cells are transplanted to the liver, but the mechanisms that underlie this process are not fully defined. We examined cyclooxygenase pathways that mediate hepatic inflammation through synthesis of prostaglandins, prostacyclins, thromboxanes, and other prostanoids following

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service