Skip to Content
Merck
All Photos(1)

Key Documents

30461

Sigma-Aldrich

DEAE-Dextran hydrochloride

BioReagent, for molecular biology

Synonym(s):

Diethylaminoethyl-dextran hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

grade

for molecular biology

product line

BioReagent

form

solid

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

solubility

H2O: 0.1 g/mL, clear

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

DEAE-Dextran, a polycationic stabilizing molecule and transfection reagent, is used in the development of sustained release protein delivery and nucleic acid transfection systems.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Tabor et al.
Current protocols in immunology, Chapter 10, Unit 10-Unit 10 (2008-04-25)
Extensive knowledge of the enzymology involved in biosynthesis and degradation of nucleic acids has permitted the development of simple methods for labeling RNA and DNA with radioisotopes or biotin. These labeled probes are used primarily for hybridization to nucleic acid
Transfection using DEAE-dextran.
Gulick T.
Current Protocols in Cell Biology, doi: 10-doi: 10 (2003)
J Zarate et al.
Journal of microencapsulation, 28(7), 614-620 (2011-07-12)
Bovine serum albumin (BSA) loaded calcium alginate microparticles (MPs) produced in this study by a w/o emulsification and external gelation method exhibited spherical and fairly smooth and porous morphology with 1.052 ± 0.057 µm modal particle size. The high permeability of the calcium
DEAE-dextran transfection of lymphocyte cell lines.
Smale ST.
Cold Spring Harbor Protocols, doi: 10-doi: 10 (2010)
Shingo Kikuchi et al.
International journal of pharmaceutics, 386(1-2), 149-155 (2009-11-20)
A multivariate statistical technique was applied to designing a tablet for the sustained release over 24h of diltiazem hydrochloride, a model highly water-soluble drug. Tablets of a hydrophilic matrix composed of dextran derivatives and hypromellose were prepared. The formulations were

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service