Skip to Content
Merck
All Photos(1)

Documents

30058

Sigma-Aldrich

Lipase Substrate

≥95% (HPLC)

Synonym(s):

1,2-Di-O-lauryl-rac-glycero-3-(glutaric acid 6-methylresorufin ester)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H69NO8
CAS Number:
Molecular Weight:
752.03
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥95% (HPLC)

form

powder

solubility

ethanol: soluble (10mg/mL)

fluorescence

λex 529 nm; λem 600 nm in 0.1 M Tris pH 8.4

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCOCC(COC(=O)CCCC(=O)Oc1ccc2N=C3C=CC(=O)C(C)=C3Oc2c1)OCCCCCCCCCCCC

InChI

1S/C45H69NO8/c1-4-6-8-10-12-14-16-18-20-22-31-50-34-38(51-32-23-21-19-17-15-13-11-9-7-5-2)35-52-43(48)25-24-26-44(49)53-37-27-28-39-42(33-37)54-45-36(3)41(47)30-29-40(45)46-39/h27-30,33,38H,4-26,31-32,34-35H2,1-3H3

InChI key

FYOMYGYHAZESGQ-UHFFFAOYSA-N

General description

Lipase substrate (1,2-Di-O-lauryl-rac-glycero-3-(glutaric,acid 6-methylresorufin) ester, DGGR) is a chromogenic substrate for lipases. It is used in colorimetric methods to measure lipase activity. This substrate produces a red-purple compound, methylresorufin, upon digestion by the lipase enzyme.

Application

Lipase Substrate is used as substrate in a high‑throughput screening assay to measure the lipase activity in response to lipstatin production

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yan Q Chen et al.
Journal of lipid research, 61(8), 1203-1220 (2020-06-04)
Angiopoietin-like protein (ANGPTL)8 has been implicated in metabolic syndrome and reported to regulate adipose FA uptake through unknown mechanisms. Here, we studied how complex formation of ANGPTL8 with ANGPTL3 or ANGPTL4 varies with feeding to regulate LPL. In human serum
Ievgeniia Ostrov et al.
Food microbiology, 82, 316-324 (2019-04-28)
Biofilm-forming Bacillus species are often involved in contamination of dairy products and therefore present a major microbiological challenge in the field of food quality and safety. In this study, we sequenced and analyzed the genomes of milk- and non-milk-derived Bacillus
Michal Híreš et al.
Current microbiology, 75(5), 580-587 (2017-12-20)
Pancreatic lipase inhibitors, such as tetrahydrolipstatin (orlistat), are used in anti-obesity treatments. Orlistat is the only anti-obesity drug approved by the European Medicines Agency (EMA). The drug is synthesized by saturation of lipstatin, a β-lactone compound, isolated from Streptomyces toxytricini
Sébastien Cadau et al.
International journal of molecular sciences, 23(21) (2022-11-12)
Atopic dermatitis (AD), the most common inflammatory skin disorder, is a multifactorial disease characterized by a genetic predisposition, epidermal barrier disruption, a strong T helper (Th) type 2 immune reaction to environmental antigens and an altered cutaneous microbiome. Microbial dysbiosis
Jose Iglesias et al.
Journal of lipid research, 57(1), 131-141 (2015-10-02)
Lipids are used as cellular building blocks and condensed energy stores and also act as signaling molecules. The glycerolipid/ fatty acid cycle, encompassing lipolysis and lipogenesis, generates many lipid signals. Reliable procedures are not available for measuring activities of several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service