Skip to Content
Merck
All Photos(1)

Documents

27815

Sigma-Aldrich

Brilliant Blue G

250, for microscopy

Synonym(s):

Acid blue 90, Coomassie Brilliant Blue G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H48N3NaO7S2
CAS Number:
Molecular Weight:
854.02
Colour Index Number:
42655
Beilstein:
5230822
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.26

grade

for microscopy

form

solid

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3C)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=CC(\C=C5C)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C47H49N3O7S2.Na/c1-6-49(31-35-11-9-13-43(29-35)58(51,52)53)40-21-25-45(33(4)27-40)47(37-15-17-38(18-16-37)48-39-19-23-42(24-20-39)57-8-3)46-26-22-41(28-34(46)5)50(7-2)32-36-12-10-14-44(30-36)59(54,55)56;/h9-30H,6-8,31-32H2,1-5H3,(H2,51,52,53,54,55,56);/q;+1/p-1

InChI key

RWVGQQGBQSJDQV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Brilliant Blue G (BBG) is a dye. It has been proposed as a substitute for internal limiting membrane (ILM) peeling.

Application

Brilliant Blue G was used to prepare the protein reagent for the determination of protein content of the collagenase enzyme isolated from fish waste. It may be employed as stain for the internal limiting membrane (ILM) for the macular hole (MH) and epiretinal membrane (ERM) surgery.

Biochem/physiol Actions

P2X7 purinergic receptor antagonist.

Analysis Note

λmax. ∼610 nm; E(1%,1 cm) ∼500-700 (H2O)

Other Notes

Protein stain for gel electrophoresis. Does not penetrate the gel and is three times more sensitive than Naphthol Blue Black B; Simple and sensitive protein assay; Rapid, sensitive and versatile assay for proteins; free amino acids and several chemicals that interfere with the Lowry procedure cause no interference.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and activation of collagenase from fish processing waste.
Daboor SM, et al.
Advances in Bioscience and Biotechnology, 3(3) (2012)
M Remy et al.
The British journal of ophthalmology, 92(8), 1142-1147 (2008-07-26)
To evaluate the retinal toxicity of Brilliant Blue G (BBG) following intravitreal injection in rat eyes and examine the biocompatibility and the staining properties in humans. BBG was injected into the 11 rat eyes to evaluate toxic effects with balanced
Hiroshi Enaida et al.
Retina (Philadelphia, Pa.), 26(6), 631-636 (2006-07-11)
To report the use of the dye brilliant blue G (BBG) for staining of the internal limiting membrane (ILM) during macular hole (MH) and epiretinal membrane (ERM) surgery. This study was designed as an interventional, noncomparative, prospective, clinical case series.
W.J. Shaphonov
Methods in Enzymology, 11, 184-184 (1967)
A rapid, sensitive, and versatile assay for protein using Coomassie brilliant blue G250.
J J Sedmak et al.
Analytical biochemistry, 79(1-2), 544-552 (1977-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service