Skip to Content
Merck
All Photos(1)

Key Documents

234A-1

Sigma-Aldrich

hCG Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (234A-14)
vial of 0.5 mL concentrate (234A-15)
bottle of 1.0 mL predilute (234A-17)
vial of 1.0 mL concentrate (234A-16)
bottle of 7.0 mL predilute (234A-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:250

control

placenta

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... NUP42(11097)

General description

Human chorionic gonadotropin (hCG) is a hormone synthesized by syncytiotrophoblasts and belongs to a group of glycoprotein hormones known as gonadotropins. Gonadotropins regulate normal growth, sexual development, and reproductive function. This antibody detects syncytiotrophoblasts in placenta and in tumors such as choriocarcinoma and some cases of seminoma.
hCG is a protein secreted in large quantities by normal trophoblasts; the antibody detects cells and tumors of trophoblastic origin such as choriocarcinoma. Large cell carcinoma and adenocarcinoma of the lung demonstrate anti-hCG positivity in 90% and 60% of cases respectively. 20% of lung squamous cell carcinomas are positive for anti-hCG. hCG expression by non-trophoblastic tumors may indicate aggressive behavior since it has been observed that hCG may play a role in the host response to a given tumor.

Quality


IVD

IVD

IVD

RUO

Linkage

hCG Positive Control Slides, Product No. 234S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Kurman et al.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists, 3(1), 101-121 (1984-01-01)
This report presents preliminary observations on the immunocytochemical localization of human chorionic gonadotropin (hCG) and human placental lactogen (hPL) in placental site trophoblastic tumors, hydatidiform moles, and choriocarcinomas and compares the findings with those of a similar immunocytochemical analysis of
L D Boucher et al.
Human pathology, 26(11), 1201-1206 (1995-11-01)
The authors studied the expression of trophoblastic cell markers in lung carcinomas cells by immunoperoxidase staining using antibodies against three trophoblastic glycoproteins (human chorionic gonadotropin [hCG]; human placental lactogen [hPL]; pregnancy-specific beta 1-glycoprotein [SP-1]). One hundred five tissue sections from
Determination of biological activity of gonadotropins hCG and FSH by Forster resonance energy transfer based biosensors.?
Mazina, Olga et al.
Scientific Reports, 7, 42219-42219 (2017)
R J Kurman et al.
Cancer, 38(6), 2404-2419 (1976-12-01)
The clinical and pathologic features of 71 endodermal sinus tumors of the ovary were studied in an effort to delineate the histogenesis and biologic behavior of this neoplasm and to evaluate the efficacy of different forms of treatment. Alpha-fetoprotein (AFP)
Choriogonadotropin positive seminoma-a clinicopathological and molecular genetic study of 15 cases.
Hes, Ondrej et al
Annals of diagnostic pathology, 18,2, 89-94 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service