Skip to Content
Merck
All Photos(1)

Documents

C1063

Sigma-Aldrich

Chorionic gonadotropin human

lyophilized powder, vial of ~2,500 IU

Synonym(s):

Choriogonin, HCG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.32

biological source

human urine

Quality Level

sterility

sterile-filtered

form

lyophilized powder

mol wt

37.9 kDa

contains

mannitol

packaging

vial of ~2,500 IU

drug control

Home Office Schedule 4.2

impurities

HBsAg, HCV and HIV-1/HIV-2, tested negative

solubility

water: 10 mL/vial, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... CGA(1081)

Looking for similar products? Visit Product Comparison Guide

General description

Glycoprotein hormone consisting of a 92-amino acid α-chain which is identical to that of LH, FSH, and TSH; and a distinct 149-amino acid β-chain. Produced by chorionic tissue and responsible for maintaining the corpus luteum during early pregnancy. Also produced by trophoblast cell neoplasms, which are marked by elevated free hCG β-chains and hyperglycosylated hCG.

Application

Chorionic gonadotropin human has been used in the biological validation of urinary testosterone (UTM) and urinary estradiol metabolites (UEM) EIA and superovulation.

Biochem/physiol Actions

Human chorionic gondadotropin (hCG) may be used as an alternative to PMSG to induce ovulation in laboratory animals. When hCG was injected into laboratory animals via intravenous (i.v.), intraperitoneal (i.p.), or subcutaneous (s.c.) methodstet, the effective dose values for 50% of the animals (ED50) per animal and per body weight (kg) in parenthesis were as follows:
Mice: i.v. 0.2 (7.7) I.U.; i.p. 0.3 (11.5) I.U.; s.c. 0.7 (26.9) I.U.
Syrian hamsters: i.v. 1.0 (9.5) I.U.; i.p. 1.8 (17.1) I.U.; s.c. 2.6 (24.8) I.U.
Rats: i.v. 1.3 (4.6) I.U.; i.p. 3.5 (12.3) I.U.; s.c.7.5 (26.3) I.U.

Using PMSG, the ED50 values per animal and per body weight (kg) in parenthesis were as follows:
Mice: i.v. 0.8 (30.8) I.U.; i.p. 2.0 (76.9) I.U.; s.c. 2.8 (107.7) I.U
Syrian hamsters: i.v., 3.6 (34.3) I.U.; i. p., 8.0 (76.2) I.U.; s.c., 13.2 (125.7) I.U.
Rats: i.v., 6.0 (76.8) I.U.; i p., 20.8 (73.0) I.U.; s.c., 76.8 (269.5) I.U.

Reconstitution

When reconstituted with water to a concentration of 1,000 IU/mL, the vial will also contain 0.1 M sodium phosphate buffer (pH approx. 7.2) and 100 mg/mL of mannitol.

Analysis Note

The calculated value is based on the USP method for human Chorionic Gonadotrophin, to meet 2,500 IU per vial, with a tolerance of not less than 80.0% and not more than 125.0% of the potency stated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interrelationship among annual cycles of sex steroids, corticosterone and body condition in Nyctibatrachus humayuni
Joshi AM, et al.
General and Comparative Endocrinology, 260, 151-160 (2018)
Luqing Zhang et al.
PloS one, 10(3), e0120396-e0120396 (2015-03-25)
ZFN, TALENs and CRISPR/Cas9 system have been used to generate point mutations and large fragment deletions and insertions in genomic modifications. CRISPR/Cas9 system is the most flexible and fast developing technology that has been extensively used to make mutations in
Katherine M Graham et al.
Conservation physiology, 6(1), coy020-coy020 (2018-05-08)
Establishing captive breeding populations of amphibians is an important conservation strategy to safeguard against ongoing declines of wild populations and provide broodstock for reintroduction programs. The endangered dusky gopher frog (DGF) has never naturally reproduced in captivity and requires breeding
Emmet L Guy et al.
Conservation physiology, 8(1), coz113-coz113 (2020-01-16)
Many amphibian species reinitiate the processes of preparing for reproduction (e.g. oogenesis) soon after breeding indicating hormone-induced ovulation could potentially be achieved out-of-season, which would lead to higher annual fecundity compared to mono-seasonal breeding. Such strategies would be beneficial to
SMN is required for the maintenance of embryonic stem cells and neuronal differentiation in mice.
Chang W F, et al.
Brain Structure &Amp; Function, 220(3), 1539-1553 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service