Skip to Content
Merck
All Photos(1)

Documents

228M-1

Sigma-Aldrich

BG8, LewisY (F3) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

F3, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (228M-14)
vial of 0.5 mL concentrate (228M-15)
bottle of 1.0 mL predilute (228M-17)
vial of 1.0 mL concentrate (228M-16)
bottle of 7.0 mL predilute (228M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:10-1:50

isotype

IgM

control

adenocarcinoma of lung

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... F3(2152)

General description

Blood group antigens have been examined as potential discriminators between pulmonary adenocarcinoma (PACA) and epithelioid mesothelioma (EM). Lewisy is the only one of these that appears to have some merit. BG8 is raised from the SK-LU-3 lung cancer line and its ability to distinguish between PACA and EM was first reported by Jordon and colleagues in 1989. Three groups have since reported their results. These studies included 231 cases of PACA and 197 cases of EM. Sensitivity and specificity for PACA were both 93%. Yaziji H et al. reported a sensitivity of nonmesothelial antigens for adenocarcinoma as organ dependent, with BG8 performing at 98% in the breast cancer group, and 100% in the lung cancer group. The specificity of the nonmesothelial (non-EM) antigens for adenocarcinoma was 98% for BG8. They concluded using logical regression analysis that a three-antibody immunohistochemical panel including calretinin, BG8, and MOC-31 would provide 96% sensitivity and specificity for distinguishing EM from adenocarcinoma from a variety of sources (lung, ovary, breast, stomach).

Quality


IVD

IVD

IVD

RUO

Linkage

BG8 Positive Control Slides, Product No. 228S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alberto M Marchevsky et al.
Applied immunohistochemistry & molecular morphology : AIMM, 15(2), 140-144 (2007-05-26)
There are no firmly established guidelines for the use of antibodies in immunohistology as individual tests or panels. Practicing pathologists must rely on information available in individual publications, review articles, books, and internet-based databases to develop diagnostic immunohistochemical algorithms for
J E King et al.
Histopathology, 48(3), 223-232 (2006-01-25)
Immunohistochemistry is frequently employed to aid the distinction between mesothelioma and pulmonary adenocarcinoma metastatic to the pleura, but there is uncertainty as to which antibodies are most useful. We analysed published data in order to establish sensitivity and specificity of
B Davidson et al.
Virchows Archiv : an international journal of pathology, 435(1), 43-49 (1999-08-04)
The detection of malignant cells in pleural, peritoneal, and pericardial fluids of cancer patients marks the presence of metastatic disease and is associated with a grave prognosis. We evaluated five epithelial markers for the detection of cancer cells in 94
N G Ordóñez
The American journal of surgical pathology, 24(4), 598-606 (2000-04-11)
The distinction between malignant pleural mesotheliomas and adenocarcinomas, particularly those originating in the lung, is a difficult diagnostic problem that can be facilitated by the use of immunohistochemical markers. In this study, the immunoreactivity of thyroid transcription factor-1 (TTF-1), E-cadherin
Nelson G Ordóñez
The American journal of surgical pathology, 27(8), 1031-1051 (2003-07-29)
A large number of immunohistochemical markers that can facilitate the distinction between epithelioid pleural mesotheliomas and pulmonary peripheral adenocarcinomas have recently become available. The aim of this study is to compare the value of these new markers with others that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service