Skip to Content
Merck
All Photos(1)

Documents

224M-1

Sigma-Aldrich

Beta-Catenin (14) Mouse Monoclonal Antibody

clone 14, unconjugated, Cell Marque

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

14, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (224M-14)
vial of 0.5 mL concentrate (224M-15)
bottle of 1.0 mL predilute (224M-17)
vial of 1.0 mL concentrate (224M-16)
bottle of 7.0 mL predilute (224M-18)

manufacturer/tradename

Cell Marque

concentration

1.0-5.0 μg/mL (predilute)
25-125 μg/mL (concentrate)

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1

control

fibromatosis of breast

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous, nuclear

General description

Beta-Catenin is a 92 kD protein normally found in the cytoplasm of the cell in the submembranous location. This protein is associated with E-Cadherin and may be essential for the function of E-Cadherin. Mutations in the Beta-Catenin gene result in nuclear accumulation of this protein. Nuclear accumulation of this protein has been demonstrated in fibromatosis lesions of the breast and abdomen and therefore is useful in differentiating this lesion from other spindle cell lesions that may occur in these locations. Nuclear accumulation of Beta-Catenin has also been demonstrated in colorectal carcinoma.

Quality


IVD

IVD

IVD

RUO

Linkage

Beta-Catenin Positive Control Slides, Product No. 224S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Li et al.
The American journal of pathology, 153(3), 709-714 (1998-09-15)
Aggressive fibromatosis is a monoclonal proliferation of spindle (fibroblast-like) cells. A subset of lesions contain somatic truncating adenomatous polyposis coli (APC) gene mutations, and all of the lesions contain an elevated beta-catenin protein level. A major function of APC is
B A Alman et al.
The American journal of pathology, 151(2), 329-334 (1997-08-01)
Sporadic aggressive fibromatosis (also called desmoid tumor) is a monoclonal proliferation of spindle (fibrocyte-like) cells that is locally invasive but does not metastasize. A similarity to abdominal fibromatoses (desmoids) in familial adenomatous polyposis and a cytogenetic study showing partial deletion
Ejaz Butt et al.
F1000Research, 8, 613-613 (2019-05-02)
Background: Ependymomas are glial tumors derived from differentiated ependymal cells. In contrast to other types of brain tumors, histological grading is not a good prognostic marker for these tumors. In order to determine genomic changes in an anaplastic ependymoma, we
Elizabeth Montgomery et al.
The American journal of surgical pathology, 26(10), 1296-1301 (2002-10-03)
Although separating gastrointestinal stromal tumor (GIST) from mesenteric fibromatosis and sclerosing mesenteritis is clinically important, this distinction sometimes poses problems for practicing pathologists. In the STI571 (Gleevec, Imatinib) era, the problem may be further compounded when protocol-driven staining for CD117
Susan C Abraham et al.
Human pathology, 33(1), 39-46 (2002-02-02)
Fibromatoses of the breast are nonmetastasizing tumors, but can be infiltrative and locally recurrent. Breast fibromatoses are rare, and their specific genetic alterations have not been elucidated. However, their occasional occurrence in patients with familial adenomatous polyposis (FAP) and their

Articles

Colorectal cancer is the third most common cancer in both men and women. An estimated 136,000 cases of colorectal cancer are expected to occur in 2016.

Related Content

Trusted IVDR-ready supplier of high-quality Cell Marque™ antibodies for diagnostic pathology and IHC, ensuring reliability and expert support.

Trusted IVDR-ready supplier of high-quality Cell Marque™ antibodies for diagnostic pathology and IHC, ensuring reliability and expert support.

Trusted IVDR-ready supplier of high-quality Cell Marque™ antibodies for diagnostic pathology and IHC, ensuring reliability and expert support.

Trusted IVDR-ready supplier of high-quality Cell Marque™ antibodies for diagnostic pathology and IHC, ensuring reliability and expert support.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service