Skip to Content
Merck
All Photos(1)

Documents

208A-7

Sigma-Aldrich

GH Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (208A-74)
vial of 0.5 mL concentrate (208A-75)
bottle of 1.0 mL predilute (208A-77)
vial of 1.0 mL concentrate (208A-76)
bottle of 7.0 mL predilute (208A-78)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

control

pituitary

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

mouse ... Gh(14599)

General description

Anti-GH is a useful marker in classification of pituitary tumors and the study of pituitary disease (acromegaly). It reacts with GH-producing cells.

Quality


IVD

IVD

IVD

RUO

Linkage

GH Positive Control Slides, Product No. 208S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kovacs et al.
Virchows Archiv. A, Pathological anatomy and histology, 395(1), 59-68 (1982-01-01)
A pituitary adenoma removed by surgery from a 22-year-old man was studied by histology, immunocytology, transmission electron microscopy and immunoelectron microscopy. Clinically, the patient had acromegaly and euthyroidism with elevated blood GH concentrations. Blood TSH and T4 levels were within
T Fukaya et al.
Cancer, 45(7), 1598-1603 (1980-04-01)
In attempting to establish a functional classification of acidophilic adenomas of the pituitary gland, immunoperoxidase staining techniques specific for growth hormone (GH), prolactin (PRL), and luteinizing hormone (LH) were carried out. Surgically removed specimens from 24 acromegalic patients were studied.
T García-Caballero et al.
Endocrine, 12(3), 265-271 (2000-08-30)
The liver is an essential target tissue for growth hormone (GH) and prolactin (PRL). The aim of this study was to determine the in situ expression of growth hormone receptor (GHR) and prolactin receptor (PRLR) in hepatocellular carcinomas and to
K S G Cunha et al.
Journal of clinical pathology, 56(10), 758-763 (2003-09-30)
The hallmark of neurofibromatosis type 1 (NF1) is the development of multiple neurofibromas. Solitary neurofibroma may occur in an individual who does not have NF1, but multiple neurofibromas tend to develop only in those with NF1. It has been suggested
L K Chopin et al.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society, 12(2), 126-136 (2002-08-15)
Prostate cancer is a significant cause of morbidity and mortality in Western males. While it is known that androgens play a central role in prostate cancer development and progression, other hormones and growth factors are also involved in prostate growth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service