Skip to Content
Merck
All Photos(1)

Key Documents

121767

Sigma-Aldrich

AG 1024

A cell-permeable, reversible, substrate competitive, and specific inhibitor of insulin-like growth factor-1 (IGF-1) and insulin receptor tyrosine kinase activity.

Synonym(s):

AG 1024, 3-Bromo-5- t-butyl-4-hydroxy-benzylidenemalonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13BrN2O
CAS Number:
Molecular Weight:
305.17
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white to yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Brc1c(c(cc(c1)C=C(C#N)C#N)C(C)(C)C)O

InChI

1S/C14H13BrN2O/c1-14(2,3)11-5-9(4-10(7-16)8-17)6-12(15)13(11)18/h4-6,18H,1-3H3

InChI key

ABBADGFSRBWENF-UHFFFAOYSA-N

General description

A cell-permeable, reversible, substrate competitive, and specific inhibitor of insulin-like growth factor-1 (IGF-1) and insulin receptor kinases with significantly lower IC50 values for IGF-1 than for insulin receptors. Induces apoptosis by downregulating the expression of phospho-Akt1 and Bcl-2; and increasing the expression of Bax, p53 and p21. Reported to enhance the radiosensitivity of human breast cancer cells.
A cell-permeable, reversible, substrate competitive, and specific inhibitor of insulin-like growth factor-1 (IGF-1) and insulin receptor tyrosine kinase activity. Exhibits lower IC50 values for IGF-1 than for the insulin receptor. Also inhibits insulin-stimulated cellular proliferation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IGF-1
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Wen, B., et al. 2001. Br. J. Cancer85, 2017.
Parrizas, M., et al. 1997. Endocrinology138, 1427.
Ohmichi, M., et al. 1993. Biochemistry32, 4650.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service