1391
Sigma-Sil-A
suitable for preparation of TMS derivatives
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.75
Recommended Products
form
liquid
Quality Level
suitability
suitable for preparation of TMS derivatives
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
Application
Trimethylchlorosilane: hexamethyldisilazane: pyridine (1:3:9) reagent for preparing TMS derivatives of amino acids, sugars, etc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
68.0 °F - closed cup
Flash Point(C)
20 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Alexander J Nedopil et al.
International orthopaedics, 41(2), 283-291 (2016-09-14)
Thirteen patients presented with patellofemoral instability out of 3212 knees treated with kinematically aligned total knee arthroplasty (KA TKA) during a nine year period. We determined the clinical characteristics and post-operative radiographic parameters associated with patellofemoral instability, and whether re-operation and
Sweeley, C.C., et al.
Journal of the American Chemical Society, 85, 2497-2497 (1963)
Yan Li et al.
Journal of cellular physiology, 235(7-8), 5811-5822 (2020-02-01)
Although we have obtained porcine pluripotent stem cell lines (pPSCs) from blastocysts, the cells exhibit flat clonal morphology and do not support single-cell passage. There is massive cell death after cell dissociation, and the efficiency of single-cell colony is generally
Abeer Maher Fayyad et al.
International journal of molecular sciences, 20(7) (2019-04-04)
Obesity is a major risk for diabetes. Brown adipose tissue (BAT) mediates production of heat while white adipose tissue (WAT) function in the storage of fat. Roles of BAT in the treatment of obesity and related disorders warrants more investigation.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service