Skip to Content
Merck
All Photos(1)

Key Documents

11702

Sigma-Aldrich

Bacitracin

from Bacillus licheniformis, ≥65 IU/mg

Synonym(s):

Altracin, Bacitracin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H103N17O16S
CAS Number:
Molecular Weight:
1422.69
EC Number:
MDL number:
UNSPSC Code:
51282002
PubChem Substance ID:
NACRES:
NA.85

biological source

Bacillus licheniformis

Quality Level

form

powder

specific activity

≥65 IU/mg

color

white to pale buff

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[H]N1[C@@H](Cc2cnc[nH]2)C(=O)N[C@H](CC(O)=O)C(=O)N[C@@H](CC(N)=O)C(=O)NCCCC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](CCC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)C3CSC(=N3)C(N)C(C)CC)[C@@H](C)CC)C(=O)N[C@H](CCCN)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@H](Cc4ccccc4)C1=O

InChI

1S/C66H103N17O16S/c1-9-35(6)52(69)66-81-48(32-100-66)63(97)76-43(26-34(4)5)59(93)74-42(22-23-50(85)86)58(92)83-53(36(7)10-2)64(98)75-40-20-15-16-25-71-55(89)46(29-49(68)84)78-62(96)47(30-51(87)88)79-61(95)45(28-39-31-70-33-72-39)77-60(94)44(27-38-18-13-12-14-19-38)80-65(99)54(37(8)11-3)82-57(91)41(21-17-24-67)73-56(40)90/h12-14,18-19,31,33-37,40-48,52-54H,9-11,15-17,20-30,32,67,69H2,1-8H3,(H2,68,84)(H,70,72)(H,71,89)(H,73,90)(H,74,93)(H,75,98)(H,76,97)(H,77,94)(H,78,96)(H,79,95)(H,80,99)(H,82,91)(H,83,92)(H,85,86)(H,87,88)/t35?,36-,37-,40-,41+,42+,43-,44+,45-,46-,47+,48?,52?,53-,54-/m0/s1

InChI key

CLKOFPXJLQSYAH-RNHDWVCBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Bacitracin is used to study disruption of bacterial cell wall synthesis at the level of peptidoglycan biosynthesis and isoprenyl metabolism. It is also used to study the biosynthesis of sterols and squalene.
Bacitracin has been used:
  • as a protease inhibitor
  • as a control antibiotic
  • for the calibration in size exclusion chromatography

Biochem/physiol Actions

Bacitracin has been shown to decrease vancomycin-resistant Enterococcus faecium population in intestine.
Bacitracin is a peptide antibiotic.
Antimicrobial spectrum: Gram-positive bacteria.
Mode of Action: Inhibits bacterial cell wall synthesis by inhibiting dephosphorylation of lipid pyrophosphate.

Packaging

5G, 25 G

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashwantha Kumar Enjapoori et al.
Genome biology and evolution, 6(10), 2754-2773 (2014-09-24)
Monotremes (platypus and echidna) are the descendants of the oldest ancestor of all extant mammals distinguished from other mammals by mode of reproduction. Monotremes lay eggs following a short gestation period and after an even briefer incubation period, altricial hatchlings
Oral Bacitracin: A Consideration for Suppression of Intestinal Vancomycin-Resistant Enterococci (VRE) and for VRE Bacteremia From an Apparent Gastrointestinal Tract Source.
Truc T Tran et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 60(11), 1726-1728 (2015-02-24)
Anthony W Kingston et al.
Molecular microbiology, 93(1), 37-49 (2014-05-09)
Heptaprenyl diphosphate (C35 -PP) is an isoprenoid intermediate in the synthesis of both menaquinone and the sesquarterpenoids. We demonstrate that inactivation of ytpB, encoding a C35 -PP utilizing enzyme required for sesquarterpenoid synthesis, leads to an increased sensitivity to bacitracin
Hiromasa Tsuda et al.
Antimicrobial agents and chemotherapy, 46(12), 3756-3764 (2002-11-19)
Streptococcus mutans is resistant to bacitracin, which is a peptide antibiotic produced by certain species of Bacillus. The purpose of this study was to clarify the bacitracin resistance mechanism of S. mutans. We cloned and sequenced two S. mutans loci
Kazuhiko Nakano et al.
Nature communications, 2, 485-485 (2011-09-29)
Although several risk factors for stroke have been identified, one-third remain unexplained. Here we show that infection with Streptococcus mutans expressing collagen-binding protein (CBP) is a potential risk factor for haemorrhagic stroke. Infection with serotype k S. mutans, but not

Articles

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service