Skip to Content
Merck
All Photos(1)

Key Documents

D5155

Sigma-Aldrich

Dihydrostreptomycin sesquisulfate

powder, suitable for cell culture, BioReagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H41N7O12 · 3/2H2SO4
CAS Number:
Molecular Weight:
730.71
Beilstein:
3894221
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.76

product name

Dihydrostreptomycin sesquisulfate, BioReagent, suitable for cell culture, ≥95.0% (HPLC)

product line

BioReagent

Quality Level

Assay

≥95.0% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.CN[C@H]1[C@H](O)[C@@H](O)[C@H](CO)O[C@H]1O[C@H]2[C@@H](O[C@@H](C)[C@]2(O)CO)O[C@@H]3[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]3NC(N)=N.CN[C@H]4[C@H](O)[C@@H](O)[C@H](CO)O[C@H]4O[C@H]5[C@@H](O[C@@H](C)[C@]5(O)CO)O[C@@H]6[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]6NC(N)=N

InChI

1S/2C21H41N7O12.3H2O4S/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4/h2*5-18,26,29-36H,3-4H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4)/t2*5-,6-,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,21+;;;/m00.../s1

InChI key

CZWJCQXZZJHHRH-YZTFXSNBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Dihydrostreptomycin sesquisulfate is a derivative of streptomycin used in aminoglyside uptake studies. Other studies have also used it as an electrode for a cochlear amplifier in the hair-cell bundle of lizards.

Biochem/physiol Actions

Mode of Action: Dihydrostreptomycin sesquisulfate inhibits protein synthesis at the level o the 30S ribosomal subunit and the 16s rRNA.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Cherlet et al.
Journal of mass spectrometry : JMS, 42(5), 647-656 (2007-04-19)
Dihydrostreptomycin (DHS) is an aminoglycoside antibiotic used in veterinary medicine in combination with benzylpenicillin for the treatment of bacterial infections in cattle, pigs and sheep. A method to determine its residues in edible tissues of cattle, as well as in
Katy Jeannot et al.
Journal of bacteriology, 187(15), 5341-5346 (2005-07-21)
MexXY is an inducible efflux system that contributes to the natural resistance of Pseudomonas aeruginosa to antibiotics. Experiments involving real-time PCR after reverse transcription in reference strain PAO1 showed concentration-dependent induction of gene mexY by various ribosome inhibitors (e.g., chloramphenicol
Y Sasaki et al.
Epidemiology and infection, 140(11), 2074-2081 (2012-01-28)
This study determined the prevalence and antimicrobial susceptibility of Salmonella isolated from broiler flocks in Japan. Caecal dropping samples were collected from 288 broiler flocks between November 2007 and February 2010. Salmonella was prevalent in 248 (86·1%) broiler flocks. The
Shin-ichi Kawano
Rapid communications in mass spectrometry : RCM, 23(6), 907-914 (2009-02-19)
Impurities in streptomycin (STR) and dihydrostreptomycin (DHS) were investigated by hydrophilic interaction chromatography/electrospray ionization quadrupole ion trap/time-of-flight mass spectrometry (HILIC/ESI-QIT/TOFMS). Samples were separated on a fused-core silica column (100 mmx2.1 mm i.d., particle size: 2.7 microm) with isocratic elution using
Murali Pendela et al.
Rapid communications in mass spectrometry : RCM, 23(12), 1856-1862 (2009-05-19)
Dihydrostreptomycin sulphate (DHS) is a water-soluble, broad-spectrum aminoglycoside antibiotic. For quantitative analysis, the European Pharmacopoeia (Ph. Eur.) prescribes an ion-pairing liquid chromatography/ultraviolet (LC/UV) method using a C18 stationary phase. Several unknown compounds were detected in commercial samples. Hence, for characterization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service