Skip to Content
Merck
All Photos(1)

Key Documents

10829

Sigma-Aldrich

trans-β-Apo-8′-carotenal

~20% apocarotenal basis (UV-vis), suspension (oily)

Synonym(s):

Apocarotenal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H40O
CAS Number:
Molecular Weight:
416.64
Beilstein:
2064131
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

biological source

corn (or maize)

form

suspension (oily)

concentration

~20% (apocarotenal, UV-vis)

color

black to brown
brown to very dark brown

mp

138-141 °C

storage temp.

2-8°C

SMILES string

[H]C(=O)\C(C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C30H40O/c1-24(13-8-9-14-25(2)16-11-18-27(4)23-31)15-10-17-26(3)20-21-29-28(5)19-12-22-30(29,6)7/h8-11,13-18,20-21,23H,12,19,22H2,1-7H3/b9-8+,15-10+,16-11+,21-20+,24-13+,25-14+,26-17+,27-18+

InChI key

DFMMVLFMMAQXHZ-DOKBYWHISA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-β-Apo-8′-carotenal is a linear aldehydic carotenoid. It is naturally found in oranges, spinach, grass, tangerines and marigolds and thus, it is directly absorbed from the diet.

Application

trans-β-Apo-8′-carotenal has been used as an internal standard during vitamin E analysis.

Biochem/physiol Actions

trans-β-Apo-8′-carotenal is commercially available as a food colorant.
Natural carotenoid that is formed from β-carotene in vivo and is an inducer of cytochrome P450 1A (CYP1A) in rats and in Ah-receptor-responsive mice.

Caution

solidifies on refrigeration

Physical form

Suspension of micronized crystals of apocarotenal dispersed in vegetable oils

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescence properties of the allenic carotenoid fucoxanthin: Analysis of the effect of keto carbonyl group by using a model compound, all-trans-beta-apo-8?-carotenal
Mimuro M, et al.
Journal of Luminescence, 51(1-3), 1-10 (1992)
Maryam Anjomshoa et al.
Stem cells and development, 18(2), 259-267 (2008-04-22)
Understanding neuroectoderm formation and its subsequent diversification to functional neural subtypes remains elusive. We have shown here for the first time that embryonic stem cells (ESCs) can differentiate into neurons and motor neurons (MNs) by using a coculture embryonic notochord
The Effects of Various Levels of Docosahexaenoic Acid on Inflammatory Markers in Conditioned Horses During Lactate Threshold Tests
Hess T, et al.
Journal of equine veterinary science, 72(6), 64-71 (2019)
M Hannuksela et al.
International journal of dermatology, 25(3), 178-180 (1986-04-01)
Double-blind, placebo controlled challenge tests with benzoic acid butylhydroxytoluene, butylhydroxyanisole, beta-carotene, beta-8-apo-carotenal, and sodium metabisulfite were made in 44 cases of chronic urticaria, 91 cases of atopic dermatitis, and 123 cases of contact dermatitis, as a comparison group. Positive reactions
A B Barua et al.
The Journal of nutrition, 130(8), 1996-2001 (2000-08-05)
beta-Carotene might be converted oxidatively to vitamin A- active products in animals by the following three possible routes: 1) central cleavage, 2) sequential excentric cleavage or 3) random cleavage. Central cleavage is strongly favored by stoichiometric studies with tissue homogenates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service